Skip to Content
Merck
All Photos(1)

Key Documents

Y0000822

Molsidomine impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Nitrosomorpholine, NMOR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8N2O2
CAS Number:
Molecular Weight:
116.12
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

molsidomine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=NN1CCOCC1

InChI

1S/C4H8N2O2/c7-5-6-1-3-8-4-2-6/h1-4H2

InChI key

ZKXDGKXYMTYWTB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Molsidomine impurity B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Song et al.
Scientific reports, 10(1), 8329-8329 (2020-05-21)
The gloomy outcome of liver cancer is mainly due to the high rates of metastasis and recurrence, even after curative resection for early stage liver cancer. Our study was conducted to find the animal model suitable for the study of
Suramya Waidyanatha et al.
Xenobiotica; the fate of foreign compounds in biological systems, 50(11), 1341-1351 (2020-06-06)
The specialty amine catalyst 2,2'-dimorpholinodiethyl ether (DMDEE) is a high-production volume chemical used in the production of flexible foam, high-resilient molded foam, and in coatings and adhesives. The disposition and metabolism of [14C]DMDEE (20 or 200 mg/kg) were determined in male
Hekap Kim et al.
Environmental health and toxicology, 26, e2011003-e2011003 (2011-11-30)
N-Nitrosodimethylamine (NDMA) is classified as a probable human carcinogen by the United States Environmental Protection Agency (US EPA) and is formed during the chlorination of municipal drinking water. In this study, selected nitrosamines were measured in chlorinated drinking water collected
Hitoshi Kodamatani et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 36(11), 1393-1399 (2020-07-28)
An ultra-sensitive analytical system that can determine the concentration of N-nitrosamines at the ng/L level without preconcentration was used to investigate the contamination, decomposition, and formation of N-nitrosodimethylamine (NDMA) and other N-nitrosamines in water samples during general analytical procedures. A
Noelia Ramírez et al.
Journal of chromatography. A, 1426, 191-200 (2015-12-20)
Thirdhand tobacco smoke (THS) constitutes a poorly understood pathway of exposure of non-smokers, especially toddlers, to tobacco-related carcinogens. However, to date most of the carcinogens present in tobacco smoke have not been detected in THS and, therefore, the significance of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service