Skip to Content
Merck
All Photos(1)

Key Documents

Y0000788

Indinavir

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Indinavir monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H47N5O4 · H2O
CAS Number:
Molecular Weight:
631.80
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

indinavir

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N4([C@@H](CN(CC4)Cc5cnccc5)C(=O)NC(C)(C)C)C[C@@H](O)C[C@@H](Cc3ccccc3)C(=O)N[C@@H]1[C@@H](Cc2c1cccc2)O.O

InChI

1S/C36H47N5O4.H2O/c1-36(2,3)39-35(45)31-24-40(22-26-12-9-15-37-21-26)16-17-41(31)23-29(42)19-28(18-25-10-5-4-6-11-25)34(44)38-33-30-14-8-7-13-27(30)20-32(33)43;/h4-15,21,28-29,31-33,42-43H,16-20,22-24H2,1-3H3,(H,38,44)(H,39,45);1H2/t28-,29+,31+,32-,33+;/m1./s1

InChI key

XTYSXGHMTNTKFH-BDEHJDMKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Indinavir EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julieta C Imperiale et al.
Drug development and industrial pharmacy, 40(12), 1607-1615 (2013-09-21)
This work investigated the production of pure indinavir free base nanoparticles by a supercritical anti-solvent method to improve the drug dissolution in intestine-like medium. To increase the dissolution of the drug by means of a supercritical fluid processing method. Acetone
Dario Akaberi et al.
Journal of biomolecular structure & dynamics, 38(18), 5526-5536 (2019-12-28)
Zika virus (ZIKV) is an emerging mosquito-borne flavivirus and infection by ZIKV Asian lineage is known to cause fetal brain anomalies and Guillain-Barrés syndrome. The WHO declared ZIKV a global public health emergency in 2016. However, currently neither vaccines nor
Oh-Kyung Kwon et al.
Virology journal, 12, 53-53 (2015-04-17)
In South Korea, about 20 types of antiretroviral drugs are used in the treatment of patients with human immunodeficiency virus/acquired immune deficiency syndrome. Since 2010, raltegravir, etravirine, and darunavir have been spotlighted as new drugs for highly active antiretroviral therapy
Camille B Blake et al.
American journal of physiology. Regulatory, integrative and comparative physiology, 307(6), R711-R720 (2014-07-06)
Pathologies in which insulin is dysregulated, including diabetes, can disrupt central vagal circuitry, leading to gastrointestinal and other autonomic dysfunction. Insulin affects whole body metabolism through central mechanisms and is transported into the brain stem dorsal motor nucleus of the
Yasuo Uchida et al.
The Journal of pharmacology and experimental therapeutics, 350(3), 578-588 (2014-06-21)
The aim of this study was to investigate whether in vivo drug distribution in brain in monkeys can be reconstructed by integrating four factors: protein expression levels of P-glycoprotein (P-gp)/multidrug resistance protein 1 at the blood-brain barrier (BBB), in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service