Skip to Content
Merck
All Photos(1)

Key Documents

SML0189

Sigma-Aldrich

Indinavir sulfate salt hydrate

≥98% (HPLC), powder, antiretroviral agent

Synonym(s):

2,3,5-Trideoxy-N-[(1S,2R)-2,3-dihydro-2-hydroxy-1H-inden-1-yl]-5-[(2S)-2-[[(1,1-dimethylethyl)amino]carbonyl]-4-(3-pyridinylmethyl)-1-piperazinyl]-2-(phenylmethyl)-D-erythro-pentonamide sulfate hydrate, Crixivan hydrate, L-735,524 hydrate, MK-639 hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H47N5O4·H2SO4 · xH2O
Molecular Weight:
711.87 (anhydrous basis)
UNSPSC Code:
51111800
NACRES:
NA.77

Product Name

Indinavir sulfate salt hydrate, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

H2O: ≥15 mg/mL

originator

Merck & Co., Inc., Kenilworth, NJ, U.S.

storage temp.

2-8°C

Application

Indinavir sulfate salt hydrate may be used in HIV-related cell signaling studies.

Biochem/physiol Actions

Indinavir sulfate is an antiretroviral; HIV Protease Inhibitor
Indinavir sulfate is an antiviral HIV protease inhibitor.
Indinavir sulfate suppresses the replication of HIV and is an important component of antiretroviral therapy for initial treatment of HIV infection. It is known to cause renal and urologic toxicity.

Features and Benefits

This compound was developed by Merck & Co., Inc., Kenilworth, NJ, U.S.. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan D Cooper et al.
Nephron. Clinical practice, 118(3), c262-c268 (2011-01-08)
The introduction of potent combination antiretroviral therapy (ART) in the treatment of HIV infection has permitted reliable control of disease progression and has markedly improved survival among people with HIV. As a result, health care providers and patients have shifted
Benjamin Seyer et al.
Journal of neurochemistry, 153(4), 485-494 (2019-09-27)
Ethyl2-acetylamino-7-hydroxy-4-pyridin-3-yl-4H-chromene-3-carboxylate (HFI-419), the benzopyran-based inhibitor of insulin-regulated aminopeptidase (IRAP), has previously been shown to improve spatial working and recognition memory in rodents. However, the mechanism of its cognitive-enhancing effect remains unknown. There is a close correlation between dendritic spine density

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service