Skip to Content
Merck
All Photos(4)

Documents

V8388

Sigma-Aldrich

Vincristine sulfate

meets USP testing specifications

Synonym(s):

Vincristine sulfate salt, 22-Oxovincaleukoblastine sulfate salt, Leurocristine sulfate salt, VCR

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H56N4O10 · H2SO4
CAS Number:
Molecular Weight:
923.04
Beilstein:
3924631
EC Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.21

Agency

USP/NF
meets USP testing specifications

Quality Level

solubility

ethanol: slightly soluble
methanol: soluble
water: freely soluble

antibiotic activity spectrum

neoplastics

application(s)

pharmaceutical (small molecule)

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

OS(O)(=O)=O.CC[C@]1(O)CC2CN(CCc3c([nH]c4ccccc34)[C@@](C2)(C(=O)OC)c5cc6c(cc5OC)N(C=O)C7[C@](O)([C@H](OC(C)=O)[C@]8(CC)C=CCN9CC[C@]67C89)C(=O)OC)C1

InChI

1S/C46H56N4O10.H2O4S/c1-7-42(55)22-28-23-45(40(53)58-5,36-30(14-18-48(24-28)25-42)29-12-9-10-13-33(29)47-36)32-20-31-34(21-35(32)57-4)50(26-51)38-44(31)16-19-49-17-11-15-43(8-2,37(44)49)39(60-27(3)52)46(38,56)41(54)59-6;1-5(2,3)4/h9-13,15,20-21,26,28,37-39,47,55-56H,7-8,14,16-19,22-25H2,1-6H3;(H2,1,2,3,4)/t28-,37-,38+,39+,42-,43+,44+,45-,46-;/m0./s1

InChI key

AQTQHPDCURKLKT-PNYVAJAMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Vincristine sulfate is an alkaloid present in Cantharanthus roseus.

Application

Vincristine sulfate has been used:
  • as a positive control in potato disc anti-tumor bioassay
  • as a standard cytotoxic agent
  • to treat cells for manipulation of the microtubule network

Widely used in cancer research. In studies on multi-drug resistance. Used to inhibit cell cycle progression at M-phase. Used to inhibit monoamine oxidase (MAO).

Biochem/physiol Actions

Vincristine sulfate possesses antitumor action targeting cell cycle. It prevents the action of tubulin by inhibiting microtubule polymerization. It also causes cell death and blocks metaphase in mitosis by preventing chromosome segregation. Vincristine sulfate inhibits angiogenesis mediated by factors such as vasculature endothelial growth factor. It decreases vascularization in tumors and normal tissues.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Muta. 2 - Repr. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytotoxicity and immunomodulating characteristics of labdane diterpenes from Marrubium cylleneum and Marrubium velutinum
Karioti A, et al.
Phytochemistry, 68(11), 1587-1594 (2007)
Molecularly characterized solvent extracts and saponins from Polygonum hydropiper L. show high anti-angiogenic, anti-tumor, brine shrimp, and fibroblast NIH/3T3 cell line cytotoxicity
Ayaz M, et al.
Frontiers in Pharmacology, 7, 74-74 (2016)
Haq Nawaz et al.
The Analyst, 138(20), 6177-6184 (2013-08-24)
Understanding the interaction of anticancer drugs with model cell lines is important to elucidate the mode of action of these drugs as well as to develop cost effective and rapid screening methods. Raman spectroscopy has been demonstrated to be a
Nicole D Paris et al.
Scientific reports, 10(1), 19501-19501 (2020-11-13)
Pediatric cancer treatment often involves chemotherapy and radiation, where off-target effects can include skeletal muscle decline. The effect of such treatments on juvenile skeletal muscle growth has yet to be investigated. We employed a small animal irradiator to administer fractionated
The polyomavirus BK agnoprotein co-localizes with lipid droplets
Unterstab G, et al.
Virology, 399(2), 322-331 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service