Skip to Content
Merck
All Photos(3)

Key Documents

T3383

Sigma-Aldrich

Tetracycline hydrochloride

≥95% (European Pharmacopoeia HPLC assay)

Synonym(s):

4-Epitetracycline hydrochloride, Achromycin hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O8 · HCl
CAS Number:
Molecular Weight:
480.90
Beilstein:
3844873
EC Number:
MDL number:
UNSPSC Code:
51284043
PubChem Substance ID:
NACRES:
NA.76
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (European Pharmacopoeia HPLC assay)

form

powder

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

color

yellow

mp

220-223 °C (lit.)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

storage temp.

−20°C

SMILES string

Cl.CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)[C@]2(O)C(=O)C(C(N)=O)=C1O)C(=O)c4c(O)cccc4[C@@]3(C)O

InChI

1S/C22H24N2O8.ClH/c1-21(31)8-5-4-6-11(25)12(8)16(26)13-9(21)7-10-15(24(2)3)17(27)14(20(23)30)19(29)22(10,32)18(13)28;/h4-6,9-10,15,25,27-28,31-32H,7H2,1-3H3,(H2,23,30);1H/t9-,10-,15-,21+,22-;/m0./s1

InChI key

XMEVHPAGJVLHIG-FMZCEJRJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: tetracycline

Application

Tetracycline is a broad spectrum polyketide antibiotic with clinical uses in treating bacterial infections such as Rocky Mountain spotted fever, typush fever, tick fevers, Q fever, and Brill-Zinsser disease and to treat upper respiratory infections and acne. It has been used in studies of multidrug resistance and potential side effects including acute pancreatitis. It is recommended for use in cell culture applications at 10 mg/L.

Biochem/physiol Actions

Mode of Action: Tetracycline passively diffuses through proin channels in the cell membrane, binding to 30S ribosomes and inhibits protein synthesis by preventing access of aminoacyl tRNA to the acceptor site on the mRNA-ribosome complex. It also binds to the bacterial 50S ribosomal subunit, altering the membrane and causing intracellular components to leak from bacterial cells. The inhibitory effects can be reversed by washing, suggesting that it is the reversibly bound antibiotic, and not the irreversibly bound drug, that is responsible for antibacterial action.

Mode of Resistance: The effects are inactivated via a loss of cell wall permeability.

Antimicrobial spectrum: Includes a wide range of antimicrobial activity against gram-positive and gram-negative bacteria.

Caution

This product should be frozen below 0°C and protected from light and moisture. In these conditions, the product has been shown to retain activity for 4 years. Stock solutions should be stored at -20°C and are stable at 37°C for 4 days.

Preparation Note

The product is freely soluble in water, soluble in methanol and ethanol but is insoluble in ether and hydrocarbons. In water, the product yields a clear, yellow-orange solution with heating and in 95% ethanol, 50 mg dissolved in 4 mL with heating yields a clear, yellow-green solution. Tetracycline is rapidly destroyed by alkali hydroxide solutions and standing water solutions become turbid due to hydrolysis and precipitation. The potency of tetracycline is reduced in solutions with pH below 2.

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Signal Word

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abulaziz Juma et al.
Frontiers in microbiology, 11, 1477-1477 (2020-08-01)
One current strategy to deal with the serious issue of antibiotic resistance is to use biosurfactants, weak antimicrobials in their own right, with antibiotics in order to extend the efficacy of antibiotics. Although an adjuvant effect has been observed, the
Rachid Elaini et al.
Insects, 11(10) (2020-10-01)
The Mediterranean fruit fly (medfly), Ceratitis capitata, is a significant pest of stone and pome fruit that causes considerable economic losses worldwide. Current control is primarily based on insecticides, which are often mixed with protein baits. Oxitec has developed a
Yuping Cao et al.
mSystems, 6(6), e0098821-e0098821 (2021-11-03)
Assembly of a resistome in parallel with the establishment of a microbial community is not well understood. Germfree models can reveal microbiota interactions and shed light on bacterial colonization and resistance development under antibiotic pressure. In this study, we exposed
Diego López-Márquez et al.
Plant methods, 16, 41-41 (2020-03-25)
Small RNAs are sequence-dependent negative regulators of gene expression involved in many relevant plant processes such as development, genome stability, or stress response. Functional characterization of sRNAs in plants typically relies on the modification of the steady state levels of
Huanhuan Jia et al.
Frontiers in cell and developmental biology, 8, 83-83 (2020-03-07)
To determine the relationship between maternal bone resorption and bone development in fetuses. Female SD rats were injected with either fluorescent calcium indicator calcein alone or together with tetracycline 1 week before pregnancy, followed by fluorescence detection in fetal tibias

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service