Skip to Content
Merck
All Photos(4)

Documents

T22500

Sigma-Aldrich

N,N,N′,N′-Tetramethylethylenediamine

ReagentPlus®, 99%

Synonym(s):

1,2-Bis(dimethylamino)ethane, TEMED, TMEDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCH2CH2N(CH3)2
CAS Number:
Molecular Weight:
116.20
Beilstein:
1732991
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Quality Level

product line

ReagentPlus®

Assay

99%

expl. lim.

9.08 %

refractive index

n20/D 1.4179 (lit.)

bp

120-122 °C (lit.)

mp

−55 °C (lit.)

density

0.775 g/mL at 20 °C (lit.)

SMILES string

CN(C)CCN(C)C

InChI

1S/C6H16N2/c1-7(2)5-6-8(3)4/h5-6H2,1-4H3

InChI key

KWYHDKDOAIKMQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N,N′,N′-Tetramethylethylenediamine may be used as an initiator along with ammonium persulfate for polymerization reactions.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

61.7 °F - closed cup

Flash Point(C)

16.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and Dynamics of Poly (N-isopropylacrylamide)? Clay Nanocomposite Gels.
Shibayama M, et al.
Macromolecules, 37(25), 9606-9612 (2004)
Teruyuki Hayashi et al.
Polymers, 11(8) (2019-08-07)
Cationic nanogels of N-isopropylacrylamide (NIPAM), including NIPAM-based cationic fluorescent nanogel thermometers, were synthesized with a cationic radical initiator previously developed in our laboratory. These cationic nanogels were characterized by transmission electron microscopy (TEM), dynamic light scattering (DLS), zeta potential measurements
Xin Ku et al.
Journal of combinatorial chemistry, 11(3), 338-340 (2009-03-06)
A practical and promising protocol was developed for S-arylations of various thiols with different substituted aryl halides. The reactions were readily facilitated to afford desired thioethers under mild conditions in good to excellent yields. The versatility, air-stability, operational simplicity of
Andrew J Ross et al.
The Journal of organic chemistry, 76(6), 1727-1734 (2011-02-16)
The structure of the alkylzinc-tetramethylethyl-enediamine (TMEDA) cluster cation 3 has been determined in the gas phase by a combination of tandem mass spectrometry, infrared multiphoton dissociation (IRMPD) spectroscopy, and DFT calculations. Both sets of experimental results establish the existence of
Jack K Clegg et al.
Dalton transactions (Cambridge, England : 2003), 39(11), 2804-2815 (2010-03-05)
New examples of nitrogen base adducts of dinuclear Co(II), Ni(II) and Cu(II) complexes of the doubly deprotonated forms of 1,3-aryl linked bis-beta-diketones of type [RC(=O)CH(2)C(=O)C(6)H(4)C(=O)CH(2)C(=O)R] (L(1)H(2)) incorporating the mono- and difunctional amine bases pyridine (Py), 4-ethylpyridine (EtPy), piperidine (pipi), 1,4-piperazine

Articles

Bis-Tris SDS-PAGE gel casting kits with mPAGE® TurboMix technology for rapid, reliable gel casting and protein separation.

Bis-Tris SDS-PAGE gel casting kits with mPAGE® TurboMix technology for rapid, reliable gel casting and protein separation.

Bis-Tris SDS-PAGE gel casting kits with mPAGE® TurboMix technology for rapid, reliable gel casting and protein separation.

Bis-Tris SDS-PAGE gel casting kits with mPAGE® TurboMix technology for rapid, reliable gel casting and protein separation.

Protocols

Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.

Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.

Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.

Simplify your SDS-PAGE gel preparation with our pre-mixed solutions and protocols to achieve precise protein separation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service