Skip to Content
Merck
All Photos(1)

Key Documents

T0700000

Theobromine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2,6-Dihydroxy-3,7-dimethylpurine, 3,7-Dihydro-3,7-dimethyl-1H-purine-2,6-dione, 3,7-Dimethylxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N4O2
CAS Number:
Molecular Weight:
180.16
Beilstein:
16464
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

theobromine

manufacturer/tradename

EDQM

mp

345-350 °C

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[H]N1C(N(C)C2=C(N(C)C=N2)C1=O)=O

InChI

1S/C7H8N4O2/c1-10-3-8-5-4(10)6(12)9-7(13)11(5)2/h3H,1-2H3,(H,9,12,13)

InChI key

YAPQBXQYLJRXSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Theobromine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Phosphosdiesterase inhibitor; weak adenosine receptor antagonist; diuretic; smooth muscle relaxant.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of theobromine, theophylline and caffeine in cocoa samples by a high-performance liquid chromatographic method with on-line sample cleanup in a switching-column system.
Brunetto R D M, et al.
Food Chemistry, 100(2), 459-467 (2007)
Analysis of caffeine, theobromine and theophylline in coffee by near infrared spectroscopy (NIRS) compared to high-performance liquid chromatography (HPLC) coupled to mass spectrometry.
Huck W C, et al.
Analytica Chimica Acta, 538(1-2), 195-203 (2005)
B Stavric
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 26(8), 725-733 (1988-08-01)
This review provides a brief overview of known information on the human toxicity of theobromine and paraxanthine. Theobromine has some pharmacological effects, although these activities are considerably weaker than those of theophylline and/or caffeine, described in parts 1 and 2
Malen Massot-Cladera et al.
The British journal of nutrition, 112(12), 1944-1954 (2014-10-28)
A diet containing 10% cocoa, a rich source of polyphenols and fibre, is able to modify intestinal immune status as well as microbiota composition. The present study was aimed at investigating whether cocoa flavonoid content is uniquely responsible for these
J Timson
Mutation research, 32(2), 169-178 (1975-01-01)
Theobromine and theophylline have a limited therapeutic use and in addition they occur in plants used in the preparation of a number of widely consumed drinks. Thus most of the population must be exposed to both compounds. Chromosome abnormalities are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service