Skip to Content
Merck
All Photos(1)

Key Documents

PHR1284

Supelco

Chlorpropamide

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

1-(p-Chlorobenzenesulfonyl)-3-propylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13ClN2O3S
CAS Number:
Molecular Weight:
276.74
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. C1905000
traceable to USP 1126009

API family

chlorpropamide

CofA

current certificate can be downloaded

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCCNC(=O)NS(=O)(=O)c1ccc(Cl)cc1

InChI

1S/C10H13ClN2O3S/c1-2-7-12-10(14)13-17(15,16)9-5-3-8(11)4-6-9/h3-6H,2,7H2,1H3,(H2,12,13,14)

InChI key

RKWGIWYCVPQPMF-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Chlorpropamide belongs to the sulfonylurea class of drugs and is found to be clinically effective in the treatment of Diabetes mellitus (DM). Its mode of action involves the stimulation of insulin release from the β-cells of the pancreas in response to a glucose load. It helps in increasing the number of insulin receptors, thereby enhancing the effect of insulin on the liver and promotes peripheral glucose utilization.

Application

Chlorpropamide may be used as a pharmaceutical reference standard for the quantification of the analyte in pharmaceutical formulations using capillary zone elctrophoresis technique and spectrophotometric technique.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA0019 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A capillary zone electrophoretic method for the determination of hypoglycemics as adulterants in herbal formulations used for the treatment of diabetes
Viana C, et al.
Analytical Methods : Advancing Methods and Applications, 5(8), 2126-2133 (2013)
Stephan P Tenbaum et al.
Nature medicine, 18(6), 892-901 (2012-05-23)
The Wnt–β-catenin and PI3K-AKT-FOXO3a pathways have a central role in cancer. AKT phosporylates FOXO3a, relocating it from the cell nucleus to the cytoplasm, an effect that is reversed by PI3K and AKT inhibitors. Simultaneous hyperactivation of the Wnt–β-catenin pathway and
Spectrophotometric Determination of Chlorpropamide in Bulk and Dosage Form by Complexation with Chloranilic Acid
Mbah JC and Okorie HN
Journal of Scientific Research, 3(1), 2126-2133 (2011)
Qing Zhu et al.
Molecular pharmaceutics, 7(4), 1291-1300 (2010-06-17)
As a result of an increase in the number of emerging therapies with dissolution limited bioavailability, formulation strategies such as solid dispersions that enhance the rate of solubilization are of interest. In this study, the microstructure of solid dispersions prepared
Yury V Seryotkin et al.
Acta crystallographica. Section B, Structural science, 69(Pt 1), 77-85 (2013-02-01)
The crystal structure of the high-pressure polymorph (α') of an antidiabetic drug, chlorpropamide [4-chloro-N-(propylaminocarbonyl)benzenesulfonamide, C(10)H(13)ClN(2)O(3)S], which is formed at ~2.8 GPa from the α-polymorph (P2(1)2(1)2(1)) on hydrostatic compression in saturated ethanol solution, has been determined. As a result of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service