Skip to Content
Merck
All Photos(1)

Key Documents

M9017

Supelco

Mepirizole

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H14N4O2
CAS Number:
Molecular Weight:
234.25
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CC1=NC(N2C(OC)=CC(C)=N2)=NC(OC)=C1

InChI

1S/C11H14N4O2/c1-7-5-9(16-3)13-11(12-7)15-10(17-4)6-8(2)14-15/h5-6H,1-4H3

InChI key

RHAXSHUQNIEUEY-UHFFFAOYSA-N

General description

Mepirizole belongs to the class of non-steroidal anti-inflammatory drugs (NSAIDs), widely used in the treatment of pain and inflammation.

Application

Mepirizole may be used as an analytical reference standard for the quantification of the analyte in animal samples using ultra-high-performance liquid chromatography with tandem mass spectrometry.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ikeda et al.
Chemical & pharmaceutical bulletin, 44(9), 1700-1706 (1996-09-01)
(1H-Pyrazol-1-yl)-, (1H-imidazol-1-yl)-, and (1H-1,2,4-triazol-1-yl)pyrimidines were prepared and evaluated for cytoprotective antiulcer activity. Among them, 4-methoxy-6-methyl-2-(1H-pyrazol-1-yl)pyrimidine (18) showed potent inhibition of the HCl-ethanol-induced and water-immersion stress-induced ulcers in rats, as well as low acute toxicity.
S Iinuma et al.
Digestive diseases and sciences, 43(8), 1657-1664 (1998-09-02)
The role of active oxygen species and lipid peroxidation in the pathogenesis of duodenal ulcers induced by mepirizole was investigated in rats. Oral administration of mepirizole (200 mg/kg) resulted in ulcer lesions in the proximal duodenum. Thiobarbituric acid-reactive substances (TBA-reactive
S Evangelista et al.
Neuroscience letters, 112(2-3), 352-355 (1990-05-04)
Substance P-like immunoreactivity (SP-li) is decreased in duodenal samples of animals treated with a single dose of an ulcerogen such as dulcerozine, mepirizole or cysteamine. Unlike dulcerozine- or mepirizole-induced ulcers the degree of cysteamine-induced duodenal lesions is inversely correlated with
D Renzi et al.
Advances in experimental medicine and biology, 298, 129-135 (1991-01-01)
We have investigated the endogenous levels of duodenal calcitonin gene-related peptide- (CGRP) and Substance P- (SP) like immunoreactivity (li) following the induction of duodenal ulcers in rats. Using three duodenal ulcerogens, namely cysteamine, dulcerozine or mepirizole given in a single
F Iwata et al.
Digestive diseases and sciences, 40(5), 1112-1117 (1995-05-01)
The hyperemia at the duodenal ulcer margin is important for ulcer healing. We studied the effect of tobacco cigarette smoke on the hyperemia at the margin of mepirizole-induced duodenal ulcer. Duodenal mucosal blood flow values measured by iodo[14C]antipyrine (IAP) autoradiography

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service