Skip to Content
Merck
All Photos(5)

Documents

I5502

Sigma-Aldrich

Isopropyl β-D-1-thiogalactopyranoside

≥99% (TLC)

Synonym(s):

IPTG, Isopropyl β-D-thiogalactoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18O5S
CAS Number:
Molecular Weight:
238.30
Beilstein:
4631
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.31

description

non-ionic

Quality Level

Assay

≥99% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

impurities

10.0-22.0% Dioxane

color

white to off-white

mp

120 -122  °C ((248 - 252 °F ))

solubility

water: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

CC(C)S[C@@H]1OC(CO)[C@H](O)C(O)[C@H]1O

InChI

1S/C9H18O5S/c1-4(2)15-9-8(13)7(12)6(11)5(3-10)14-9/h4-13H,3H2,1-2H3/t5-,6+,7+,8-,9+/m1/s1

InChI key

BPHPUYQFMNQIOC-NXRLNHOXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

IPTG is commonly used in cloning procedures that require induction of β-galactosidase activity. It is used in conjunction with X-Gal or Bluo-Gal in blue-white selection of recombinant bacterial colonies that induce expression of the lac operon in Escherichia coli. IPTG functions by binding to the lacI repressor and altering its conformation, which prevents the repression of the β-galactosidase coding gene lacZ.

Other Notes

Non-metabolizable galactose analog.
To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Reconstitution

A stock solution (0.1 M) is prepared by dissolving IPTG in water with subsequent sterile filtration of the solution. The final concentration of IPTG in indicator plates should be 0.2 mM.

Signal Word

WarningDanger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2

Supplementary Hazards

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iltaf Ahmed et al.
Human molecular genetics, 24(11), 3172-3180 (2015-02-24)
There are two known mRNA degradation pathways, 3' to 5' and 5' to 3'. We identified likely pathogenic variants in two genes involved in these two pathways in individuals with intellectual disability. In a large family with multiple branches, we
Therese Wohlschlager et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(27), E2787-E2796 (2014-06-01)
Effector proteins of innate immune systems recognize specific non-self epitopes. Tectonins are a family of β-propeller lectins conserved from bacteria to mammals that have been shown to bind bacterial lipopolysaccharide (LPS). We present experimental evidence that two Tectonins of fungal
Farshid Hadifar et al.
PloS one, 9(10), e108420-e108420 (2014-10-21)
Killed avian influenza virus (AIV) vaccines have been used to control H5N1 infections in countries where the virus is endemic. Distinguishing vaccinated from naturally infected birds (DIVA) in such situations however, has become a major challenge. Recently, we introduced the
B K Kishore Reddy et al.
Antimicrobial agents and chemotherapy, 58(6), 3312-3326 (2014-04-02)
Pantothenate kinase (PanK) catalyzes the phosphorylation of pantothenate, the first committed and rate-limiting step toward coenzyme A (CoA) biosynthesis. In our earlier reports, we had established that the type I isoform encoded by the coaA gene is an essential pantothenate
Tia DiTommaso et al.
PLoS genetics, 10(10), e1004706-e1004706 (2014-10-24)
Keratins are cytoskeletal intermediate filament proteins that are increasingly being recognised for their diverse cellular functions. Here we report the consequences of germ line inactivation of Keratin 76 (Krt76) in mice. Homozygous disruption of this epidermally expressed gene causes neonatal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service