Skip to Content
Merck
All Photos(1)

Key Documents

I0600000

Isoprenaline sulfate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Isoprenaline sulfate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H36N2O10S · 2H2O
CAS Number:
Molecular Weight:
556.62
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

isoprenaline

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O)O.N(C(C)C)CC(O)c2cc(c(cc2)O)O.N(C(C)C)CC(O)c1cc(c(cc1)O)O.O.O

InChI

1S/2C11H17NO3.H2O4S.2H2O/c2*1-7(2)12-6-11(15)8-3-4-9(13)10(14)5-8;1-5(2,3)4;;/h2*3-5,7,11-15H,6H2,1-2H3;(H2,1,2,3,4);2*1H2

InChI key

CUQPTVCVZLUXJB-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isoprenaline sulfate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keyvan Yousefi et al.
Indian journal of experimental biology, 51(8), 653-660 (2013-11-16)
Isoproterenol injection (100 mg/kg; sc) produced changes in ECG pattern including ST-segment elevation and suppressed R-amplitude. The methanolic extract of M. vulgare at doses of 10, 20, and 40 mg/kg significantly amended the ECG changes. A severe myocardial necrosis and
George King et al.
American journal of physiology. Lung cellular and molecular physiology, 306(3), L292-L298 (2013-12-10)
Glucocorticoids induce lung fibroblasts to produce fibroblast-pneumocyte factor, a peptide that stimulates type II cells to synthesize pulmonary surfactant. This effect is known to be more apparent in cells derived from female fetuses, a characteristic that has been attributed to
Yafei Zhang et al.
Nan fang yi ke da xue xue bao = Journal of Southern Medical University, 33(10), 1517-1520 (2013-10-23)
To observe the changes of surface ECG and cell couplings between sinoatrial node cells and myocardial cells following transplantion of pedicled autologous sinoatrial node tissue graft into the right ventricle of a canine model of complete atrioventricular block. Ten healthy
Ilka Mathar et al.
Circulation research, 114(2), 283-294 (2013-11-15)
The Trpm4 gene has recently been associated with several disorders, including cardiac conduction diseases and Brugada syndrome. Transient receptor potential member 4 (TRPM4) proteins constitute Ca2+ -activated, but Ca2+ -impermeable, nonselective cation channels and are expressed both in atrial and
Masanobu Kawai et al.
The Journal of biological chemistry, 289(3), 1457-1466 (2013-12-05)
The circadian clock network is well known to link food intake and metabolic outputs. Phosphorus is a pivotal nutritional factor involved in energy and skeletal metabolisms and possesses a circadian profile in the circulation; however, the precise mechanisms whereby phosphate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service