Skip to Content
Merck
All Photos(1)

Key Documents

I0599990

Isoprenaline hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Isoprenaline hydrochloride, (±)-Isoproterenol hydrochloride, 1-(3′,4′-Dihydroxyphenyl)-2-isopropylaminoethanol hydrochloride, N-Isopropyl-DL-noradrenaline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO3 · HCl
CAS Number:
Molecular Weight:
247.72
Beilstein:
3917363
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

isoprenaline

manufacturer/tradename

EDQM

mp

165-175 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.CC(C)NCC(O)c1ccc(O)c(O)c1

InChI

1S/C11H17NO3.ClH/c1-7(2)12-6-11(15)8-3-4-9(13)10(14)5-8;/h3-5,7,11-15H,6H2,1-2H3;1H

InChI key

IROWCYIEJAOFOW-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Isoprenaline hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

β-adrenoceptor agonist; increases cytosolic cAMP.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Kubuš et al.
Circulation. Arrhythmia and electrophysiology, 7(2), 218-223 (2014-02-04)
Data on the results and clinical effect of an invasive risk stratification strategy in asymptomatic young patients with the Wolff-Parkinson-White electrocardiographic pattern are scarce. Eighty-five consecutive patients aged<18 years with a Wolff-Parkinson-White pattern and persistent preexcitation at maximum exercise undergoing
Inbal Rachmin et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(16), 5902-5907 (2014-04-09)
ErbB2 interacting protein (Erbin) is a widely expressed protein and participates in inhibition of several intracellular signaling pathways. Its mRNA has been found to be present in relatively high levels in the heart. However, its physiological role in the heart
Marianne Tare et al.
Clinical science (London, England : 1979), 127(9), 559-569 (2014-05-13)
Intrauterine inflammation is a major contributor to preterm birth and has adverse effects on preterm neonatal cardiovascular physiology. Cardiomyocyte maturation occurs in late gestation in species such as humans and sheep. We tested the hypothesis that intrauterine inflammation has deleterious
Chrishan S Samuel et al.
Hypertension (Dallas, Tex. : 1979), 64(2), 315-322 (2014-05-29)
Relaxin is a naturally occurring peptide hormone that mediates systemic hemodynamic and renal adaptive changes during pregnancy and abrogates aberrant scar tissue formation (fibrosis) in diverse pathogeneses. However, its efficacy relative to renin–angiotensin system blockade, the most effective antifibrotic strategy
Christiane Neuber et al.
The Journal of pharmacology and experimental therapeutics, 349(1), 39-46 (2014-01-17)
Stimulation of myocardial β(1)-adrenoceptors (AR) is a major mechanism that increases cardiac function. We investigated the functional consequences of genetic β(1)-AR knockdown in three-dimensional engineered heart tissue (EHT). For β(1)-AR knockdown, short interfering RNA (siRNA) sequences targeting specifically the β(1)-AR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service