Skip to Content
Merck
All Photos(3)

Key Documents

H0252

Sigma-Aldrich

Hydroxyapatite

Type I, buffered aqueous suspension

Synonym(s):

Calcium phosphate hydroxide, Durapatite, Hydroxylapatite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3Ca3(PO4)2 · Ca(OH)2
CAS Number:
Molecular Weight:
502.31
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
grade:
for analytical purposes
form:
buffered aqueous suspension

grade

for analytical purposes

Quality Level

type

Type I

form

buffered aqueous suspension

storage temp.

2-8°C

SMILES string

[Ca++].[Ca++].[Ca++].[Ca++].O[Ca+].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/5Ca.3H3O4P.H2O/c;;;;;3*1-5(2,3)4;/h;;;;;3*(H3,1,2,3,4);1H2/q5*+2;;;;/p-10

InChI key

XYJRXVWERLGGKC-UHFFFAOYSA-D

Looking for similar products? Visit Product Comparison Guide

Application

Useful in the purification of proteins and nucleic acids

Packaging

Package size based on dry weight

Physical form

Suspension in 0.001 M phosphate buffer, pH 6.8; approx. 25% solids

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie E P New et al.
Circulation research, 113(1), 72-77 (2013-04-26)
We previously showed that early calcification of atherosclerotic plaques associates with macrophage accumulation. Chronic renal disease and mineral imbalance accelerate calcification and the subsequent release of matrix vesicles (MVs), precursors of microcalcification. We tested the hypothesis that macrophage-derived MVs contribute
Bjarke Viberg et al.
Acta orthopaedica, 84(3), 254-259 (2013-04-19)
Elderly patients with displaced femoral neck fractures are commonly treated with a hemiarthroplasty (HA), but little is known about the long-term failure of this treatment. We compared reoperation rates for patients aged at least 75 years with displaced femoral neck
Sergio Bertazzo et al.
Nature materials, 12(6), 576-583 (2013-04-23)
The accumulation of calcified material in cardiovascular tissue is thought to involve cytochemical, extracellular matrix and systemic signals; however, its precise composition and nanoscale architecture remain largely unexplored. Using nano-analytical electron microscopy techniques, we examined valves, aortae and coronary arteries
Ehud Segal et al.
PloS one, 4(4), e5233-e5233 (2009-04-22)
There is an immense clinical need for novel therapeutics for the treatment of angiogenesis-dependent calcified neoplasms such as osteosarcomas and bone metastases. We developed a new therapeutic strategy to target bone metastases and calcified neoplasms using combined polymer-bound angiogenesis inhibitors.
H S Alghamdi et al.
Journal of dental research, 92(11), 982-988 (2013-09-24)
Osteoporotic conditions are anticipated to affect the osseointegration of dental implants. This study aimed to evaluate the effect of a radiofrequent magnetron-sputtered calcium phosphate (CaP) coating on dental implant integration upon installment in the femoral condyles of both healthy and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service