Skip to Content
Merck
All Photos(1)

Key Documents

21223

Sigma-Aldrich

Hydroxyapatite

purum p.a., ≥90% (as Ca3(PO4)2, KT)

Synonym(s):

Calcium phosphate hydroxide, Durapatite, Hydroxylapatite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
3Ca3(PO4)2 · Ca(OH)2
CAS Number:
Molecular Weight:
502.31
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

grade

purum p.a.

Quality Level

Assay

≥90% (as Ca3(PO4)2, KT)

form

powder

impurities

≤5% water (KFT)

loss

≤10% loss on ignition, 800 °C

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤2000 mg/kg

cation traces

Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cu: ≤20 mg/kg
Fe: ≤400 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤10 mg/kg

SMILES string

[Ca++].[Ca++].[Ca++].[Ca++].O[Ca+].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/5Ca.3H3O4P.H2O/c;;;;;3*1-5(2,3)4;/h;;;;;3*(H3,1,2,3,4);1H2/q5*+2;;;;/p-10

InChI key

XYJRXVWERLGGKC-UHFFFAOYSA-D

Looking for similar products? Visit Product Comparison Guide

General description

Bone and tooth implant materials have been prepared from polycrystalline hydroxyapatite. The compressive, flexural, torsional and dynamic torsional strengths of polycrystalline hydroxyapatite were investigated. Electrophoretic deposition of hydroxyapatite on to flat titanium plate material has been studied.
Hydroxyapatite is a bioceramic mainly found in minerals of bone and teeth. Due to its high biocompatibility and similarity with inorganic components found in hard tissues, it is mainly used to produce biocomposites and prostheses for bones and teeth replacement.

Application

Hydroxyapatite may be used in the preparation of bioceramic specimens by incorporation of a small amount of silicon (0.4wt%) into the structure of hydroxyapatite. It may be used for the preparation of porous microstructures, having important prosthetic applications.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hydroxyapatite synthesis and the benefits of its blend with calcium aluminate cement.
Oliveira IR, et al.
Ceramics International, 42(2), 2542-2549 (2016)
P Ducheyne et al.
Biomaterials, 7(2), 97-103 (1986-03-01)
Hydroxyapatite from two sources was electrophoretically deposited onto flat titanium plate material. Depending upon the deposition conditions various changes in the structure of the ceramic were identified. A well-adhering Ti-P compound was present at the interface. Hydroxyapatite oxygenated to various
Mechanical properties of sintered hydroxyapatite for prosthetic applications.
Akao M ,et al.
J. Mater. Sci., 16(3), 809-812 (1981)
Chemical characterization of silicon-substituted hydroxyapatite
Gibson I, et al.
Journal of Biomedical Materials Research, 44(4), 422-428 (1999)
Bjarke Viberg et al.
Acta orthopaedica, 84(3), 254-259 (2013-04-19)
Elderly patients with displaced femoral neck fractures are commonly treated with a hemiarthroplasty (HA), but little is known about the long-term failure of this treatment. We compared reoperation rates for patients aged at least 75 years with displaced femoral neck

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service