Skip to Content
Merck
All Photos(1)

Key Documents

E2503004

Etomidate impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Methyl 1-[(1RS)-1-phenylethyl]-1H-imidazole-5-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14N2O2
CAS Number:
Molecular Weight:
230.26
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

etomidate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[n]2(cncc2C(=O)OC)C(C)c1ccccc1

InChI

1S/C13H14N2O2/c1-10(11-6-4-3-5-7-11)15-9-14-8-12(15)13(16)17-2/h3-10H,1-2H3

InChI key

FHFZEKYDSVTYLL-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Etomidate impurity B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grayson A Doss et al.
Journal of zoo and wildlife medicine : official publication of the American Association of Zoo Veterinarians, 45(1), 53-59 (2014-04-10)
Metomidate hydrochloride is an imidazole-based, nonbarbiturate hypnotic drug primarily used as an immersion sedation and anesthetic agent in freshwater and marine finfish. To the authors' knowledge, there is no documentation in the literature of its use in amphibians. In this
Kenneth B Davis et al.
Comparative biochemistry and physiology. Toxicology & pharmacology : CBP, 143(1), 134-139 (2006-02-10)
Channel catfish and sunshine bass were exposed to a low-water stress event and allowed to recover in fresh water or a solution of metomidate (dl-1-(1-phenylethyl)-5-(metoxycarbonyl) imidazole hydrochloride), which inhibits the synthesis of cortisol. Change in time of plasma cortisol was
M K Hansen et al.
Journal of veterinary pharmacology and therapeutics, 26(2), 95-103 (2003-04-02)
Metomidate was administered to halibut (Hippoglossus hippoglossus) and turbot (Scophthalmus maximus) intravenously at a dose of 3 mg/kg bodyweight, as a bath treatment at a dose of 9 mg/L water for 5 min to study the disposition of metomidate, and
Maria Erlandsson et al.
Nuclear medicine and biology, 36(4), 435-445 (2009-05-09)
Two- and one-step syntheses of (18)F-labelled analogues of metomidate, such as 2-[(18)F]fluoroethyl 1-[(1R)-1-phenylethyl]-1H-imidazole-5-carboxylate (1), 2-[(18)F]fluoroethyl 1-[(1R)-1-(4-chlorophenyl)ethyl]-1H-imidazole-5-carboxylate (2), 2-[(18)F]fluoroethyl 1-[(1R)-1-(4-bromophenyl)ethyl]-1H-imidazole-5-carboxylate (3), 3-[(18)F]fluoropropyl 1-[(1R)-1-(4-bromophenyl)ethyl]-1H-imidazole-5-carboxylate (4) and 3-[(18)F]fluoropropyl 1-[(1R)-1-phenylethyl]-1H-imidazole-5-carboxylate (5) are presented. Analogues 1-5 were prepared by a two-step reaction sequence that started
Barbro Eriksson et al.
Annals of the New York Academy of Sciences, 970, 159-169 (2002-10-17)
Positron emission tomography (PET) supplies a range of labeled compounds to be used for the characterization of tumor biochemistry. Some of these have proved to be of value for clinical diagnosis, treatment follow up, and clinical research. The first routinely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service