Skip to Content
Merck
All Photos(1)

Key Documents

Y0000483

Hyoscine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(−)-Scopolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H21NO4
CAS Number:
Molecular Weight:
303.35
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

hyoscine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N1([C@@H]2C3OC3[C@H]1CC(C2)OC(=O)[C@H](CO)c4ccccc4)C

InChI

1S/C17H21NO4/c1-18-13-7-11(8-14(18)16-15(13)22-16)21-17(20)12(9-19)10-5-3-2-4-6-10/h2-6,11-16,19H,7-9H2,1H3/t11?,12-,13-,14+,15?,16?/m1/s1

InChI key

STECJAGHUSJQJN-MKXJHAPZSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hyoscine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maura L Furey et al.
JAMA psychiatry, 70(3), 280-290 (2013-02-01)
The need for improved treatment options for patients with major depressive disorder (MDD) is critical. Faster-acting antidepressants and biomarkers that predict clinical response will facilitate treatment. Scopolamine produces rapid antidepressant effects and thus offers the opportunity to characterize potential biomarkers
Eun-Jin Kim et al.
Journal of ethnopharmacology, 146(1), 294-299 (2013-01-15)
Panax ginseng (family Araliaceae) is traditionally used as a remedy for cancer, inflammation, stress and aging. To explore whether ginsenosides Rg5 and Rh3, the main constituents of heat-processed ginseng (the root of Panax ginseng), could protect memory deficit. We isolated
Young Jin Jang et al.
Behavioural brain research, 245, 113-119 (2013-02-19)
Several human studies have reported that coffee consumption improves cognitive performance. In the present study, we investigated whether instant decaffeinated coffee also ameliorates cognitive performance and attenuates the detrimental effects of scopolamine on memory. Memory performance was evaluated in Morris
Jin-Di Li et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 38(11), 1719-1724 (2013-09-10)
Transgenic Atropa belladonna with high levels of scopolamine was developed by metabolic engineering. A functional gene involved in the rate limiting enzyme of h6h involved in the biosynthetic pathway of scopolamine was over expressed in A. belladonna via Agrobacterium-mediation. The
Matthew J Kogut et al.
Journal of vascular and interventional radiology : JVIR, 24(3), 326-331 (2013-02-06)
To evaluate the impact of prophylactic use of dexamethasone and scopolamine on analgesic and antiemetic agent requirements after transarterial chemoembolization. A total of 148 patients underwent 316 rounds of chemoembolization for hepatocellular carcinoma at a single institution over a 17-month

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service