Skip to Content
Merck
All Photos(1)

Key Documents

D2920000

Disopyramide

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

α-Diisopropylaminoethyl-α-phenylpyridine-2-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H29N3O
CAS Number:
Molecular Weight:
339.47
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

disopyramide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)N(CCC(C(N)=O)(c1ccccc1)c2ccccn2)C(C)C

InChI

1S/C21H29N3O/c1-16(2)24(17(3)4)15-13-21(20(22)25,18-10-6-5-7-11-18)19-12-8-9-14-23-19/h5-12,14,16-17H,13,15H2,1-4H3,(H2,22,25)

InChI key

UVTNFZQICZKOEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Disopyramide EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Class IA antiarrhythmic; sodium channel blocker

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Tokudome et al.
Angiology, 50(4), 331-335 (1999-05-04)
The effect of intravenous administration of disopyramide (total dose 100 mg, bolus 20 mg every 5 minutes) was compared with that of propranolol (total dose 10 mg, bolus 2 mg every 5 minutes) in a patient with hypertrophic obstructive cardiomyopathy.
Procainamide and disopyramide.
C Ribeiro et al.
European heart journal, 8 Suppl A, 11-19 (1987-03-01)
J P Fauchier et al.
Archives des maladies du coeur et des vaisseaux, 84(4), 553-560 (1991-04-01)
Moricizine chlorhydrate (Ethmozine), a relatively unknown antiarrhythmic agent in France, is a derivative of Phenothiazine, related to the Vaughan-Williams Class IB drugs. A randomised, double-blind, crossover trial with Disopyramide 600 mg/day after a placebo period in 10 patients with ventricular
Pharmacokinetics and pharmacodynamics of disopyramide.
J Bonde
Danish medical bulletin, 38(2), 121-133 (1991-04-01)
L Sathyavagiswaran
Journal of forensic sciences, 32(6), 1813-1818 (1987-11-01)
Disopyramide is an oral antiarrhythmic drug which reduces conduction velocity, prolongs duration of action potential and the effective refractory period, and exerts vagolytic properties. The drug is usually well absorbed orally. The principal use of the drug is to suppress

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service