Skip to Content
Merck
All Photos(1)

Key Documents

D0734000

Dextran 10 for calibration

Dextran 10 European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

biological source

bacterial (Leuconostoc mesenteroides)

grade

pharmaceutical primary standard

API family

dextran

analyte chemical class(es)

oligosaccharides

manufacturer/tradename

EDQM

application(s)

food and beverages
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dextran 10 for calibration EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M H Hemmelder et al.
The Journal of laboratory and clinical medicine, 132(5), 390-403 (1998-11-21)
Fractional dextran clearances have been extensively used to study glomerular size selectivity. We report on an analysis of different laboratory procedures involved in measuring fractional dextran clearances. The deproteinization of plasma samples by 20% trichloroacetic acid (TCA) revealed a protein
Ana Luisa Miranda-Vilela et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(4), 3391-3403 (2013-12-03)
Dextran-functionalized maghemite fluid (DexMF) has been tested to treat Ehrlich-solid-tumor-bearing mice, evidencing its potential use in mediating magnetohyperthermia in breast cancer treatment. However, although magnetic nanoparticles tend to accumulate in tumor tissues, part of the nanomaterial can reach the blood
[HES, dextran and gelatin--indications and tolerance].
H Laubenthal et al.
Anasthesiologie, Intensivmedizin, Notfallmedizin, Schmerztherapie : AINS, 33(4), 251-255 (1998-06-09)
Mary E Pease et al.
Investigative ophthalmology & visual science, 55(4), 2564-2573 (2014-02-22)
To determine differences in scleral permeability, as measured by diffusion of macromolecules, by using fluorescence recovery after photobleaching (FRAP), with reference to differences by mouse strain, scleral region, and the effect of experimental glaucoma. In three mouse strains (B6, CD1
J Salamon et al.
RoFo : Fortschritte auf dem Gebiete der Rontgenstrahlen und der Nuklearmedizin, 186(4), 367-376 (2014-04-01)
The aim of this study was to establish co-labeling of mesenchymal stromal cells (MSC) for the detection of single MSC in-vivo by MRI and histological validation. Mouse MSC were co-labeled with fluorescent iron oxide micro-particles and carboxyfluorescein succinimidyl ester (CFSE).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service