Skip to Content
Merck
All Photos(1)

Documents

31416

Supelco

Dextran from Leuconostoc mesenteroides

analytical standard, for GPC, Mw 1,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O5)n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.24

grade

analytical standard
for GPC

Quality Level

mol wt

Mn ~1,000
Mp ~1,100
Mw ~1,300

analyte chemical class(es)

oligosaccharides

technique(s)

gel permeation chromatography (GPC): suitable

Mw/Mn

~1.30

application(s)

food and beverages

format

neat

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dextran is a polysaccharide having predominantly α-1,6-glucosidic linkages, which is produced by the bacterial species, Leuconostoc mesenteroides, which grow exclusively on sucrose containing media.
Dextran is a polysaccharide having predominantly α-1,6-glucosidic linkages, which is produced by the bacterial species, Leuconostoc mesenteroides, which grow exclusively on sucrose containing media.
Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, with a linear backbone of α-linked D-glucopyranosyl repeating units. Dextrans are found as bacterial extracellular polysaccharides. They are synthesized from sucrose by Leuconostoc mesenteroides and Lactobacillus brevis. Bacteria employ dextran in biofilm formation or as protective coatings to evade host phagocytes in the case of pathogenic bacteria.

Application

Dextran from Leuconostoc mesenteroides (Mw: 1000), has been used as an analytical standard to calibrate the column for gel permeation chromatography (GPC) for the analysis of the carbohydrate.
Use of dextrans as long and hydrophilic spacer arms improves the performance of immobilized proteins acting on macromolecules.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Research into technologies of obtaining grape must in the production of sparkling base wines.
Tkachenko O, et al.
Eastern-European Journal of Enterprise Technologies (2017)
Chemistry and Processing of Sugarbeet and Sugarcane, 9 (2013)
Chemistry and Processing of Sugarbeet and Sugarcane, 9 (2013)
Preparation and characterization of dextran from Leuconostoc mesenteroides.
Jeans RA, et al.
The Journal of Biological Chemistry, 176, 603-615 (1948)
Ana Luisa Miranda-Vilela et al.
Journal of biomedical nanotechnology, 9(7), 1261-1271 (2013-08-06)
This work aimed to test a dextran-functionalized magnetic fluid (DexMF) sample in mediating magnetohyperthermia to treat an advanced clinical Ehrlich-solid-tumor, to verify the effects of oral antioxidant administration of pequi-oil on this treatment and to investigate the potential of these

Related Content

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service