Skip to Content
Merck
All Photos(1)

Documents

D0710000

Dexamethasone acetate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Dexamethasone 21-acetate, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione-21-acetate, 9α-Fluoro-16α-methylprednisolone-21-acetate, Prednisolone F acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H31FO6
CAS Number:
Molecular Weight:
434.50
Beilstein:
2342608
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

dexamethasone

packaging

pkg of 150 mg

manufacturer/tradename

EDQM

storage condition

protect from light

color

white

solubility

water: <0.1 g/L

application(s)

pharmaceutical (small molecule)

format

neat

shipped in

ambient

storage temp.

2-8°C

SMILES string

[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1(F)[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@@H](C)[C@]4(O)C(=O)COC(C)=O

InChI

1S/C24H31FO6/c1-13-9-18-17-6-5-15-10-16(27)7-8-21(15,3)23(17,25)19(28)11-22(18,4)24(13,30)20(29)12-31-14(2)26/h7-8,10,13,17-19,28,30H,5-6,9,11-12H2,1-4H3/t13-,17+,18+,19+,21+,22+,23+,24+/m1/s1

InChI key

AKUJBENLRBOFTD-RPRRAYFGSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

Dexamethasone acetate (Decadron-LA) belongs to the class of long-acting glucocorticoids, commonly used as a pharmaceutical ingredient in creams, ointments, microemulsions, lotions, aerosols, etc.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dexamethasone acetate is used as an EP reference standard to quantify the analyte in pharmaceutical formulations using chromatography ttechniques.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dexamethasone acetate
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 10(3), 4963-4965 (2021)
Textbook of Veterinary Internal Medicine, 87-92 (2009)
Development and validation of HPLC method for analysis of dexamethasone acetate in microemulsions.
Urban MCC, et al.
Brazilian Journal of Pharmaceutical Sciences , 45(1), 87-92 (2009)
Nicoleta Butoescu et al.
Journal of microencapsulation, 25(5), 339-350 (2008-05-10)
Intra-articular drug delivery systems still suffer from too short-lasting effects. Magnetic particles retained in the joint using an external magnetic field might prolong the local release of an anti-inflammatory drug. For the purpose, superparamagnetic iron oxide nanoparticles (SPIONs) and dexamethasone
Jianfeng Xu et al.
Journal of ocular pharmacology and therapeutics : the official journal of the Association for Ocular Pharmacology and Therapeutics, 23(6), 527-540 (2007-11-16)
The aims of this study were to investigate the inhibitory efficacy of intravitreal dexamethasone acetate (DA)-loaded by (D, L-lactide-co-glycolide) (PLGA) nanoparticles on choroidal neovascularization (CNV) in a laser-induced rat model and to evaluate its mode of drug release. DA loaded

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service