Skip to Content
Merck
All Photos(1)

Documents

C9231

Sigma-Aldrich

Carbenicillin disodium

meets USP testing specifications

Synonym(s):

Carbenicillin disodium salt, α-Carboxybenzylpenicillin disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H16N2Na2O6S
CAS Number:
Molecular Weight:
422.36
Beilstein:
5722128
EC Number:
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.76

Agency

USP/NF
meets USP testing specifications

Quality Level

form

solid

solubility

H2O: 50 mg/mL

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

pharmaceutical (small molecule)

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)c3ccccc3)C(=O)N2[C@H]1C([O-])=O

InChI

1S/C17H18N2O6S.2Na/c1-17(2)11(16(24)25)19-13(21)10(14(19)26-17)18-12(20)9(15(22)23)8-6-4-3-5-7-8;;/h3-7,9-11,14H,1-2H3,(H,18,20)(H,22,23)(H,24,25);;/q;2*+1/p-2/t9?,10-,11+,14-;;/m1../s1

InChI key

RTYJTGSCYUUYAL-YCAHSCEMSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Used for selection of ampr transformed cells. Used to study the role of penicillin-sensitive transpeptidases in cell wall biosynthesis.

Biochem/physiol Actions

Mode of Action: Carboxypenicillin antibiotic that inhibits bacterial cell-wall synthesis (peptidoglycan cross-linking) by inactivating transpeptidases on the inner surface of the bacterial cell membrane.


Antimicrobial spectrum: Active against Gram-positive and Gram-negative bacteria

Analysis Note

Stable at 37 °C for 3 days

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward Avilés et al.
Organic letters, 12(22), 5290-5293 (2010-10-23)
Monamphilectine A (1), a new diterpenoid β-lactam alkaloid showing potent antimalarial activity, was isolated in milligram quantities following bioassay-directed extraction of a Puerto Rican marine sponge Hymeniacidon sp. Its structure, established by interpretation of spectral data, was confirmed unequivocally by
Steven D Brown et al.
Biochemistry, 48(47), 11155-11157 (2009-10-24)
Bacteriophage Qbeta coat protein forms uniform virus-like particles when expressed recombinantly in a variety of organisms. We have inserted the IgG-binding Z domain at the carboxy terminus of the coat protein and coexpressed this chimeric subunit with native coat protein
J Steinmann et al.
Euro surveillance : bulletin Europeen sur les maladies transmissibles = European communicable disease bulletin, 16(33) (2011-08-30)
We describe the epidemiology and characteristics of the pathogen and patients (n=7) associated with an outbreak of a carbapenem-resistant Klebsiella pneumoniae (CRKP) strain in a German university hospital from July 2010 to January 2011. Species identification and detection of carbapenem
Yasser M Abdelrahman et al.
Nucleic acids research, 39(5), 1843-1854 (2010-11-06)
Chlamydia trachomatis is an obligate intracellular bacterium that exhibits a unique biphasic developmental cycle that can be disrupted by growth in the presence of IFN-γ and β-lactams, giving rise to an abnormal growth state termed persistence. Here we have examined
Jun Yang et al.
Plant science : an international journal of experimental plant biology, 181(6), 701-711 (2011-10-01)
Efficient Agrobacterium tumefaciens-mediated transformation was developed using embryogenic suspension cell cultures of elite sweet potato (Ipomoea batatas [L.] Lam.) cultivars, including Ayamurasaki, Sushu2, Sushu9, Sushu11, Wanshu1, Xushu18 and Xushu22. Embryogenic suspension cultures were established in LCP medium using embryogenic calli

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service