Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbenicillin disodium
Synonym(s):
Carbenicillin disodium salt, α-Carboxybenzylpenicillin disodium salt
Empirical Formula (Hill Notation):
C
17
H
16
N
2
Na
2
O
6
S
CAS Number:
4800-94-6
Molecular Weight:
422.36
EC Number:
225-360-8
Products
Product Category
antibiotics (1)
Brand
Sigma-Aldrich (1)
Feature
Formula Weight
Physical Form
Activity Spectrum
Agency
Quality Segment
Application
Mode of Action
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
C9231
Carbenicillin disodium,
meets USP testing specifications
Expand
View Pricing