Skip to Content
Merck
All Photos(1)

Key Documents

BP557

4′-Isobutylacetophenone

British Pharmacopoeia (BP) Reference Standard

Synonym(s):

1-(4-Isobutylphenyl)ethanone, 1-Acetyl-4-isobutylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16O
CAS Number:
Molecular Weight:
176.25
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ibuprofen

form

liquid

quality

British Pharmacopoeia (BP) Reference Standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

BP

application(s)

pharmaceutical
pharmaceutical small molecule

format

neat

storage temp.

2-8°C

SMILES string

O=C(C)c1ccc(cc1)CC(C)C

InChI

1S/C12H16O/c1-9(2)8-11-4-6-12(7-5-11)10(3)13/h4-7,9H,8H2,1-3H3

InChI key

KEAGRYYGYWZVPC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

4′-Isobutylacetophenone BP Reference standard, intended for use in laboratory tests only as specifically prescribed in the British Pharmacopoeia.

Packaging

Unit quantity: 100 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity please visit British Pharmacopoeia

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

286.7 °F - closed cup

Flash Point(C)

141.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Explore our extensive catalogue of 700+ BP chemical reference substances for precise qualitative and quantitative testing in accordance with British Pharmacopeia compendial monographs.

Explore our extensive catalogue of 700+ BP chemical reference substances for precise qualitative and quantitative testing in accordance with British Pharmacopeia compendial monographs.

Explore our extensive catalogue of 700+ BP chemical reference substances for precise qualitative and quantitative testing in accordance with British Pharmacopeia compendial monographs.

Explore our extensive catalogue of 700+ BP chemical reference substances for precise qualitative and quantitative testing in accordance with British Pharmacopeia compendial monographs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service