Skip to Content
Merck
All Photos(1)

Key Documents

62248

Supelco

2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic acid

pharmaceutical impurity standard

Synonym(s):

1-Hydroxyibuprofen, Ibuprofen impurity L (PhEur), 2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18O3
CAS Number:
Molecular Weight:
222.28
Beilstein:
22307543
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)C(O)c1ccc(cc1)C(C)C(O)=O

InChI

1S/C13H18O3/c1-8(2)12(14)11-6-4-10(5-7-11)9(3)13(15)16/h4-9,12,14H,1-3H3,(H,15,16)

InChI key

RMOQYHYFRKTDRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chloé S Baron et al.
Cell, 179(2), 527-542 (2019-10-05)
Much of current molecular and cell biology research relies on the ability to purify cell types by fluorescence-activated cell sorting (FACS). FACS typically relies on the ability to label cell types of interest with antibodies or fluorescent transgenic constructs. However
Chi-Yeh Chung et al.
Cell reports, 29(2), 495-510 (2019-10-10)
Technological improvements enable single-cell epigenetic analyses of organ development. We reasoned that high-resolution single-cell chromatin accessibility mapping would provide needed insight into the epigenetic reprogramming and transcriptional regulators involved in normal mammary gland development. Here, we provide a single-cell resource
Dobeen Hwang et al.
Cell chemical biology, 26(9), 1229-1239 (2019-06-25)
Homogeneous antibody-drug conjugates (ADCs) that use a highly reactive buried lysine (Lys) residue embedded in a dual variable domain (DVD)-IgG1 format can be assembled with high precision and efficiency under mild conditions. Here we show that replacing the Lys with
Tanner O Monroe et al.
Developmental cell, 48(6), 765-779 (2019-02-19)
Specialized adult somatic cells, such as cardiomyocytes (CMs), are highly differentiated with poor renewal capacity, an integral reason underlying organ failure in disease and aging. Among the least renewable cells in the human body, CMs renew approximately 1% annually. Consistent
Lu Wei et al.
Molecular cell, 80(1), 140-155 (2020-10-03)
The tissue-specific deployment of highly extended neural 3' UTR isoforms, generated by alternative polyadenylation (APA), is a broad and conserved feature of metazoan genomes. However, the factors and mechanisms that control neural APA isoforms are not well understood. Here, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service