Skip to Content
Merck
All Photos(1)

Key Documents

BCR296

2,2′,3,4,4′,5′-Hexachlorobiphenyl (IUPAC No. 138)

BCR®, certified reference material

Synonym(s):

2,2′,3,4,4′,5′-Hexachlorobiphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4Cl6
CAS Number:
Molecular Weight:
360.88
Beilstein:
2508233
Ballschmiter Number:
138
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

neat

storage temp.

2-8°C

SMILES string

Clc1ccc(c(Cl)c1Cl)-c2cc(Cl)c(Cl)cc2Cl

InChI

1S/C12H4Cl6/c13-7-2-1-5(11(17)12(7)18)6-3-9(15)10(16)4-8(6)14/h1-4H

InChI key

RPUMZMSNLZHIGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR296

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

O Roots et al.
Chemosphere, 35(5), 979-985 (1997-09-23)
A comparison of PCB (polychlorinated biphenyls) contents of biota in different areas of the Baltic Sea suggests the northern part of the Gulf of Riga and Väinameri Sea (Estonian coastal water) as reference area for the Baltic Sea. The concentrations
Somiranjan Ghosh et al.
Environment international, 37(5), 838-857 (2011-04-08)
Several reports have indicated that low level of polychlorinated biphenyl (PCB) exposure can adversely affect a multitude of physiological disorders and diseases in in vitro, in vivo, and as reported in epidemiological studies. This investigation is focused on the possible
L Gribaldo et al.
Journal of toxicology and environmental health. Part A, 55(2), 121-131 (1998-10-07)
The effects of two substituted polychlorinated biphenyls, the 3,4,5,3',4,5' (PCB-169) and the 2,3,4,2',4',5' (PCB-138) forms, were examined on the expression of c-myc, c-jun, c-ras, and jun-b in 3T3-L1 cells. Northern blot analysis demonstrated that the two PCBs, which exhibit a
Linlong Yu et al.
Chemosphere, 90(9), 2347-2351 (2012-11-13)
Sensitized photodechlorination of polychlorinated biphenyl, PCB 138, in three different surfactant solutions was studied. The sensitizer of choice was leuco-methylene blue, which was produced in situ from methylene blue using either triethylamine or sodium borohydride. Three types of surfactants, anionic
Roberta Campagna et al.
Toxicological sciences : an official journal of the Society of Toxicology, 123(1), 170-179 (2011-06-16)
Developmental exposure to polychlorinated biphenyls (PCBs) has been associated with cognitive deficits in humans and laboratory animals by mechanisms that remain unknown. Recently, it has been shown that developmental exposure to 2,2',3,4,4',5'-hexachlorobiphenyl (PCB138), a food-relevant PCB congener, decreases the learning

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service