All Photos(3)
About This Item
Linear Formula:
H3BO3
CAS Number:
Molecular Weight:
61.83
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
vapor pressure
2.6 mmHg ( 20 °C)
Quality Level
Assay
≥95% (titration)
form
tablet
pH
5.1 (25 °C, 1.8 g/L)
mp
160 °C (dec.) (lit.)
solubility
water: 1 tablet/100mL, clear, colorless
storage temp.
room temp
SMILES string
OB(O)O
InChI
1S/BH3O3/c2-1(3)4/h2-4H
InChI key
KGBXLFKZBHKPEV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- Effect of Boron Content in LiOH Solutions on the Corrosion Behavior of Zr-Sn-Nb Alloy.: This study investigates how varying boron content in lithium hydroxide solutions affects the corrosion behavior of Zr-Sn-Nb alloys, highlighting the potential implications for materials used in nuclear reactors (Zhao et al., 2024).
- Evaluation of living bacterial therapy assisted by pH/reactive oxygen species dual-responsive sodium alginate-based hydrogel for wound infections.: This research evaluates a boric acid-containing hydrogel for its effectiveness in treating wound infections through pH and reactive oxygen species responsiveness, indicating innovative applications in wound care (Miao et al., 2024).
Reconstitution
Dissolve 1 tablet in 100 ml deionized water
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Hye Ryeo Lee et al.
Journal of chromatography. A, 1346, 117-122 (2014-05-09)
In liquid phase microextraction, high enrichment factors can be obtained using an acceptor phase of small volume. By hanging an acceptor drop at the separation capillary tip, single drop microextraction (SDME) can be in-line coupled with capillary electrophoresis (CE). The
Akie Shimotohno et al.
Plant & cell physiology, 56(4), 620-630 (2015-02-12)
Boron, an essential micronutrient, is transported in roots of Arabidopsis thaliana mainly by two different types of transporters, BORs and NIPs (nodulin26-like intrinsic proteins). Both are plasma membrane localized, but have distinct transport properties and patterns of cell type-specific accumulation
Albert Serrà et al.
ACS nano, 8(5), 4630-4639 (2014-05-03)
Electrodeposition from microemulsions using ionic liquids is revealed as a green method for synthesizing magnetic alloyed nanoparticles, avoiding the use of aggressive reducing agents. Microemulsions containing droplets of aqueous solution (electrolytic solution containing Pt(IV) and Co(II) ions) in an ionic
Sheng Tang et al.
Journal of chromatography. A, 1373, 31-39 (2014-12-02)
Layered double oxide hollow spheres (LDO-HSs) were synthesized and employed as a dissolvable sorbent in dispersive solid-phase extraction (DSPE) to extract eleven United States Environmental Protection Agency's priority phenols from aqueous samples. With their higher specific surface area, LDO-HSs showed
Jing Li et al.
American journal of physiology. Cell physiology, 306(10), C943-C960 (2014-03-22)
The human solute carrier (SLC26) family of anion transporters consists of 10 members (SLCA1-11, SLCA10 being a pseudogene) that encode membrane proteins containing ~12 transmembrane (TM) segments with putative N-glycosylation sites (-NXS/T-) in extracellular loops and a COOH-terminal cytosolic STAS
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service