Skip to Content
Merck
All Photos(3)

Key Documents

B7901

Sigma-Aldrich

Boric acid

Boric acid
1 of 1 reviewers received a sample product or took part in a promotion

suitable for electrophoresis, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H3BO3
CAS Number:
Molecular Weight:
61.83
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
38120104
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

grade

electrophoresis grade

vapor pressure

2.6 mmHg ( 20 °C)

Assay

≥99.5%

form

powder

technique(s)

electrophoresis: suitable

impurities

≤0.005% Insoluble in methanol

pH

8.2-10.1

mp

160 °C (dec.) (lit.)

solubility

H2O: 40 mg/mL, clear, colorless

density

1.43 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.01%

cation traces

Ca: ≤0.005%
Fe: ≤0.001%
heavy metals (as Pb): ≤0.001%

suitability

suitable for electrophoresis (as a reagent)

storage temp.

room temp

SMILES string

OB(O)O

InChI

1S/BH3O3/c2-1(3)4/h2-4H

InChI key

KGBXLFKZBHKPEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Boric acid is a weak acid and is widely used in multi-purpose solutions (MPS) as a buffering agent.[1] It is less toxic and non-volatile in nature.[2]

Application

Boric acid has been used as a component of tris borate buffer:
  • for native polyacrylamide gel electrophoresis (PAGE) and denaturing urea-PAGE of tRNA samples(80)
  • for urea-polyacrylamide electrophoresis of RNA samples(81)
  • for agarose gel electrophoresis of DNA samples(82)

Biochem/physiol Actions

Boric acid is used in the periodontal therapy as an irrigation solution as it elicits bactericidal effects in microbial biofilms in root canal. Boric acid may favor osteoblastic activity and inhibit bone loss.[2] It inhibits Candida albicans fungal infection and has potential to treat vaginal infection.[3]

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antifungal mechanisms supporting boric acid therapy of Candida vaginitis
De Seta F, et al.
The Journal of Antimicrobial Chemotherapy, 63(2), 325-336 (2008)
Nonclinical safety evaluation of boric acid and a novel borate-buffered contact lens multi-purpose solution, Biotrue? multi-purpose solution
Lehmann DM, et al.
Contact Lens & Anterior Eye : the Journal of the British Contact Lens Association, 33, S24-S32 (2010)
Antibacterial effect of different concentration of boric acid against enterococcus faecalis biofilms in root canal
Zan R, et al.
Marmara Dental Journal, 2, 76-80 (2013)
Albert Serrà et al.
ACS nano, 8(5), 4630-4639 (2014-05-03)
Electrodeposition from microemulsions using ionic liquids is revealed as a green method for synthesizing magnetic alloyed nanoparticles, avoiding the use of aggressive reducing agents. Microemulsions containing droplets of aqueous solution (electrolytic solution containing Pt(IV) and Co(II) ions) in an ionic
Sheng Tang et al.
Journal of chromatography. A, 1373, 31-39 (2014-12-02)
Layered double oxide hollow spheres (LDO-HSs) were synthesized and employed as a dissolvable sorbent in dispersive solid-phase extraction (DSPE) to extract eleven United States Environmental Protection Agency's priority phenols from aqueous samples. With their higher specific surface area, LDO-HSs showed

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service