Skip to Content
Merck
All Photos(1)

Documents

95462

Supelco

1,2,3,4-Tetrahydronaphthalene

analytical standard

Synonym(s):

Tetralin solvent

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12
CAS Number:
Molecular Weight:
132.20
Beilstein:
1446407
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.55 (vs air)

vapor pressure

0.18 mmHg ( 20 °C)

Assay

≥99.5% (GC)

autoignition temp.

723 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

0.8 %, 100 °F
5 %, 150 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.541 (lit.)
n20/D 1.542

bp

207 °C (lit.)

mp

−35 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

C1CCc2ccccc2C1

InChI

1S/C10H12/c1-2-6-10-8-4-3-7-9(10)5-1/h1-2,5-6H,3-4,7-8H2

InChI key

CXWXQJXEFPUFDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,2,3,4-Tetrahydronaphthalene may be used as a solvent in studying the fabrication of solution-processed 6,13-bis(triisopropylsilylethynyl) (TIPS) pentacene thin-film transistors with a cross-linked poly-4-vinylphenol (PVP) dielectric on a polyethersulphone (PES) substrate.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Solution-processed 6, 13-bis (triisopropylsilylethynyl)(TIPS) pentacene thin-film transistors with a polymer dielectric on a flexible substrate.
Shin SI, et al.
Semiconductor Science and Technology, 23(8), 085009-085009 (2008)
Michael A Brodney et al.
Bioorganic & medicinal chemistry letters, 21(9), 2637-2640 (2011-01-29)
A novel series of tetralin containing amino imidazoles, derived from modification of the corresponding phenyl acetic acid derivatives is described. Replacement of the amide led to identification of a potent series of tetralin-amino imidazoles with robust central efficacy. The reduction
Satish Chandra Puri et al.
Journal of biotechnology, 122(4), 494-510 (2005-12-27)
The aryl tetralin lignans are synthesized by Podophyllum sps. and are in great demand worldwide due to their use in synthesis of topoisomerase inhibitors. However, the sustained production of these aryl tetralin lignans requires large-scale harvesting from the natural environments
Laurent Piccolo et al.
ChemSusChem, 5(9), 1717-1723 (2012-09-06)
The development of cleaner fuels from conventional resources requires the finding of new hydrotreatment processes able to improve the combustion performances of fuels and limit undesirable emissions. In the context of gas oil upgrading by selective ring opening, we have
Laura Tomás-Gallardo et al.
Applied and environmental microbiology, 78(15), 5460-5462 (2012-05-29)
Glucose catabolite repression of tetralin catabolic genes in Rhodococcus sp. strain TFB was shown to be exerted by a protein homologous to transcriptional regulators of the cyclic AMP receptor (CRP)-FNR family. The protein was detected bound to putative CRP-like boxes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service