427772
Polyethylene
average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Synonym(s):
PE
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
H(CH2CH2)nH
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
powder
mol wt
average Mn ~1,700 by GPC
average Mw ~4,000 by GPC
viscosity
1.5 poise(25 °C, Brookfield Thermosel)(lit.)
mp
92 °C
acid number
<0.05 mg KOH/g
transition temp
softening point 106 °C (ring and ball)
density
0.92 g/mL at 25 °C
SMILES string
C=C
InChI
1S/C2H4/c1-2/h1-2H2
InChI key
VGGSQFUCUMXWEO-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Mold release additive, lubricant in rubber processing, extrusion and calendering aid for PVC and dispersing aid for color concentrates.PEG-4000 may be used as a precipitant during protein fractionation and virus precipitation.Influence of PEG of various molecular weights on mammalian cell hybridization.
Features and Benefits
Low molecular weight and viscosity for easy processing.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
A method for the estimation of polyethylene glycol in plasma protein fractions, Analytical Biochemistry
Sims GEC and Snape TJ
Analytical Biochemistry, 107(1), 60-63 (1980)
Reduced cortical neurotransmitPolyethylene glycol-induced mammalian cell hybridization: Effect of polyethylene glycol molecular weight and concentration.
Davidson RL, et al.
Somatic Cell Genetics, 2(3), 271-280 (1976)
Lisa K McCarthy et al.
Pediatrics, 132(1), e135-e141 (2013-06-19)
Hypothermia on admission to the NICU is associated with increased mortality in preterm infants. Many newborns are hypothermic on admission despite using polyethylene bags (PBs). Using exothermic mattresses (EMs) in addition to PBs may reduce hypothermia but increase hyperthermia. We
Melody Yee-Man Wong et al.
Rapid communications in mass spectrometry : RCM, 27(6), 713-721 (2013-02-19)
Despite various porous materials having been widely adopted as spraying tips for direct sample analysis using electrospray ionization mass spectrometry (ESI-MS), the effect of surface property and porosity of spraying tip materials on their analytical performances is not clear. Investigation
Agnieszka Karbownik et al.
Acta poloniae pharmaceutica, 70(2), 301-307 (2013-04-26)
The study concerned the stability of calcium folinate in concentrate in glass vials and diluted in polyethylene (PE) bags stored at 15-25 degrees C and 2-8 degrees C for up to 34 days. Original vials of calcium folinate injection (10
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service