Skip to Content
Merck
All Photos(1)

Documents

95401

Supelco

Menthone

analytical standard

Synonym(s):

2-Isopropyl-5-methylcyclohexanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
774527
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥81% (GC)
≥97.0% (mixture of isomers, GC)

optical activity

[α]/D -7.0±5°, neat

shelf life

limited shelf life, expiry date on the label

concentration

≤19% (isomenthone, minor component, GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.449-1.451

bp

85-88 °C/12 mmHg (lit.)

density

0.896 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(C)C1CCC(C)CC1=O

InChI

1S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-9H,4-6H2,1-3H3

InChI key

NFLGAXVYCFJBMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Menthone belongs to the group of terpenes, found in a variety of volatile oils including pennyroyal, peppermint, and geranium. It finds applications in perfume and flavor compositions. Menthone is reported as a characteristic marker for the consumption of peppermint liqueurs, herbal, and bitter liqueurs.

Application

Menthone may be used as an analytical reference standard for the quantification of the analyte in food, pharmaceutical products, serum samples and Menthae Herba using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: mentha

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Resp. Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

156.2 °F - closed cup

Flash Point(C)

69 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous Determination of (-)-Menthone and (-)-Menthol in Menthae Herba by Gas Chromatography and Principal Component Analysis
Kim HS, et al.
Natural Product Sciences, 16(3), 180-184 (2010)
Headspace solid-phase microextraction--gas chromatography--mass spectrometry determination of the characteristic flavourings menthone, isomenthone, neomenthol and menthol in serum samples with and without enzymatic cleavage to validate post-offence alcohol drinking claims
Schulz K, et al.
Analytica Chimica Acta, 646(1-2), 128-140 (2009)
Determination of menthol and menthone in food and pharmaceutical products by solid-phase microextraction--gas chromatography
Ligor M and Buszewski B
Journal of Chromatography A, 847(1-2), 161-169 (1999)
Vivian Tullio et al.
Molecules (Basel, Switzerland), 24(17) (2019-09-01)
The promising antimicrobial activity of essential oils (EOs) has led researchers to use them in combination with antimicrobial drugs in order to reduce drug toxicity, side effects, and resistance to single agents. Mentha x piperita, known worldwide as "Mentha of
Guanying Hu et al.
International immunopharmacology, 24(2), 191-197 (2014-12-17)
Danshu capsule (DSC) is a medicinal compound in traditional Chinese medicine (TCM). It is commonly used for the treatment of acute & chronic cholecystitis as well as choleithiasis. To study its choleretic effect, healthy rats were randomly divided into DSC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service