Skip to Content
Merck
All Photos(2)

Documents

93938

Supelco

Aloe-emodin

analytical standard

Synonym(s):

1,8-Dihydroxy 3-hydroxymethylanthraquinone, 1,8-Dihydroxy-3-(hydroxymethyl)anthraquinone, 3-Hydroxymethylchrysazine, Rhabarberone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
2059062
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

light orange to dark orange

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

OCc1cc(O)c2C(=O)c3c(O)cccc3C(=O)c2c1

InChI

1S/C15H10O5/c16-6-7-4-9-13(11(18)5-7)15(20)12-8(14(9)19)2-1-3-10(12)17/h1-5,16-18H,6H2

InChI key

YDQWDHRMZQUTBA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aloe-emodin is a hydroxyanthraquinone and an active component present in Aloe vera leaves and root/rhizome of Rheum palmatum, possessing specific in vitro and in vivo antineuroectodermal tumor activity.

Application

Aloe-emodin may be used as a reference standard in the determination of aloe-emodin in Aloe vera extracts and commercial formulations using high-performance liquid chromatography (HPLC) coupled with tandem UV absorption and fluorimetric detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Laxative/cathartic compound; increases the contraction of intestinal smooth muscle by releasing endogenous acetylcholine. Anti-tumor activity is associated with an increased production of reactive oxygen species (ROS) that, in turn, reduces the mitochondrial transmembrane electrical potential, thus inducing a permeability transition that sets in motion a series of events culminating in cellular apoptosis. Genotoxicity and mutagenicity appear to be due to the inhibition of topoisomerase II activity by aloe emodin.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: aloe

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of aloe emodin in Aloe vera extracts and commercial formulations by HPLC with tandem UV absorption and fluorescence detection.
Mandrioli R, et al.
Food Chemistry, 126(1), 387-393 (2011)
Aloe-emodin is a new type of anticancer agent with selective activity against neuroectodermal tumors.
Pecere T, et al.
Cancer Research, 60(11), 2800-2804 (2000)
Effects and mechanisms of aloe-emodin on cell death in human lung squamous cell carcinoma.
Lee Z-H, et al.
European Journal of Pharmacology, 431(3), 287-295 (2001)
Naraganahalli R Thimmegowda et al.
Chemical biology & drug design, 85(5), 638-644 (2014-10-18)
In this study, we have synthesized novel water soluble derivatives of natural compound aloe emodin 4(a-j) by coupling with various amino acid esters and substituted aromatic amines, in an attempt to improve the anticancer activity and to explore the structure-activity
Timurs Maculins et al.
Cell chemical biology, 27(11), 1441-1451 (2020-07-30)
Protein-protein interactions (PPIs) govern intracellular life, and identification of PPI inhibitors is challenging. Roadblocks in assay development stemming from weak binding affinities of natural PPIs impede progress in this field. We postulated that enhancing binding affinity of natural PPIs via

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service