Skip to Content
Merck
All Photos(1)

Key Documents

90836

Millipore

Fraser Supplement

suitable for microbiology

Synonym(s):

Amonium Iron(III) Citrate Supplement

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41171614
NACRES:
NA.74

sterility

sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages
pathogen testing

microbiology

suitability

Listeria spp.

SMILES string

[Fe]21OC(=O)C(CC(=O)O2)(CC(=O)O1)O.N

InChI

1S/C6H8O7.Fe.H3N/c7-3(8)1-6(13,5(11)12)2-4(9)10;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;1H3/q;+3;/p-3

InChI key

FRHBOQMZUOWXQL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Recommended by ISO Committee for selective isolation and enumeration of Listeria monocytogenes from food, animal feeds etc.

Application

for selective isolation and enumeration of Listeria monocytogenes from food, animal feeds etc.

Components

(per vial, sufficient for 500 ml medium)
Ferric ammonium citrate 250 mg

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alejandro Miguel Figueroa-López et al.
Brazilian journal of microbiology : [publication of the Brazilian Society for Microbiology], 50(3), 817-824 (2019-04-13)
This study focuses on the prevalence of Listeria monocytogenes (Lm) in pork meat and on inert surfaces from slaughterhouses in Sonora, Mexico. A total of 21 Lm were obtained from 103 samples, giving a prevalence of 20.3%. The prevalence of
Jirapas Sripetchwandee et al.
Life sciences, 92(4-5), 298-304 (2013-01-22)
Although iron overload induces oxidative stress and brain mitochondrial dysfunction, and is associated with neurodegenerative diseases, brain mitochondrial iron uptake has not been investigated. We determined the role of mitochondrial calcium uniporter (MCU) in brain mitochondria as a major route
Guo-Yang Zhao et al.
Biological trace element research, 150(1-3), 487-495 (2012-10-12)
Bone metabolism has a close relationship with iron homeostasis. To examine the effects of iron excess and iron deficiency on the biological activities of osteoblast in vitro, human osteoblast cells (hFOB1.19) were incubated in a medium supplemented with 0-200 μmol/L ferric
Gabriela A Salvador et al.
Neurotoxicology, 32(1), 75-82 (2010-12-07)
Excessive neuronal iron has been proposed to contribute to the pathology of several neurodegenerative diseases including Alzheimer's and Parkinson's diseases. This work characterized human neuroblastoma IMR-32 cells exposure to ferric ammonium citrate (FAC) as a model of neuronal iron overload
Satoru Morita et al.
Japanese journal of radiology, 27(10), 423-429 (2009-12-26)
To compare three-dimensional segmented true fast imaging with steady-state precession magnetic resonance cholangiopancreatography (3D-trueFISP-MRCP) to conventional MRCP sequences with an oral negative contrast agent for diagnosing juxtapapillary diverticulum. A total of 42 patients with (n = 21) and without (n

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service