Skip to Content
Merck

RES20400-A7

Ferric ammonium citrate

Synonym(s):

Ammonium iron(III) citrate, Ammonium ferric citrate, Ferric ammonium citrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H8O7 ·xFe3+ · yNH3
CAS Number:
NACRES:
NA.21
UNSPSC Code:
41171614
EC Number:
214-686-6
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Level

form

powder

impurities

Trace metals; tested

suitability

suitable for manufacturing use

SMILES string

N.OC12CC(=O)O[Fe](OC(=O)C1)OC(=O)C2

InChI

1S/C6H8O7.Fe.H3N/c7-3(8)1-6(13,5(11)12)2-4(9)10;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;1H3/q;+3;/p-3

InChI key

FRHBOQMZUOWXQL-UHFFFAOYSA-K

General description

Our SAFC® portfolio of high-quality raw materials for use in biopharmaceutical processing withstands strict quality control procedures plus the documentation and expertise to help our customers meet requirements as defined by the M-Clarity Program.

M-Clarity Program

Our comprehensive portfolio of upstream process chemicals not only provides biopharmaceutical manufacturers with high-quality raw materials for production of classical and novel therapies, but also helps them get to market faster and simplify regulatory challenges. Trust us to deliver supply chain transparency and reliable sourcing around the globe, streamlining your product qualification with best-in-class regulatory support and service.
To request documentation for this product, please contact Customer Support and select ‘Product Documentation′. Please note that access to documentation for this product requires a confidentiality disclosure agreement.

Packaging

Product is available in the following package sizes:
RES20400-A702X: 1 kg container
RES20400-A703X: 5 kg container

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

13 - Non Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



T Takahara et al.
Nihon rinsho. Japanese journal of clinical medicine, 56(11), 2865-2869 (1998-12-16)
The high signal intensity from the intestinal fluid usually deteriorates the projection images of MR cholangiopancreatography (MRCP) because it superimposes on the biliary tract. FerriSeltz (Otsuka pharmaceutical, Tokushima, Japan; a ferric ammonium citrate-based positive oral contrast agent) solution in a
Danyu Li et al.
British journal of pharmacology, 178(5), 1182-1199 (2021-01-16)
Acute kidney injury is a common clinical problem with no definitive or specific treatment. Therefore, the molecular mechanisms of acute kidney injury must be fully understood to develop novel treatments. Nuciferine, a major bioactive compound isolated from the lotus leaf



Global Trade Item Number

SKUGTIN
RES20400-A702X04061826006603
RES20400-A703X04061832610610