Skip to Content
Merck
All Photos(1)

Documents

89050

Sigma-Aldrich

Thiosemicarbazide

puriss. p.a., 98%

Synonym(s):

Thiocarbamoyl hydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CSNHNH2
CAS Number:
Molecular Weight:
91.14
Beilstein:
506320
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

puriss. p.a.

Quality Level

Assay

98%

form

crystals

ign. residue

≤0.08% (as SO4)

mp

180-183 °C (dec.) (lit.)

anion traces

sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

NNC(N)=S

InChI

1S/CH5N3S/c2-1(5)4-3/h3H2,(H3,2,4,5)

InChI key

BRWIZMBXBAOCCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thiosemicarbazide is an environmentally toxic compound, which is usually derived from thiourea. Its ability to chelate trace metals promotes its biological activity against certain tumors, protozoa, influenza, pesticides, and fungicides. It is widely used as a metal complexing agent in various fields involving the characterization of aliphatic or aromatic aldehydes, ketones, and polysaccharides.

Application

Thiosemicarbazide has been used as an analytical reagent for the measurement of urea net high-affinity uptake in roots of intact maize plants using colorimetric assay.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Chronic 3

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical determination of thiosemicarbazide using the glassy carbon electrode modified with multi-walled carbon nanotubes
Faizbakhsh N and Safari Z
Advances in Nanochemistry, 1(2), 52-55 (2019)
Measurement of net high-affinity urea uptake in maize plants
Zanin L, et al.
Bio-protocol, 5 (2015)
Pedro I da S Maia et al.
Inorganic chemistry, 51(3), 1604-1613 (2012-01-12)
Na[AuCl(4)]·2H(2)O reacts with tridentate thiosemicarbazide ligands, H(2)L1, derived from N-[N',N'-dialkylamino(thiocarbonyl)]benzimidoyl chloride and thiosemicarbazides under formation of air-stable, green [AuCl(L1)] complexes. The organic ligands coordinate in a planar SNS coordination mode. Small amounts of gold(I) complexes of the composition [AuCl(L3)] are
Agata Siwek et al.
Journal of molecular modeling, 19(3), 1319-1324 (2012-11-29)
The preliminary cytotoxic effect of 4-ethoxycarbonylmethyl-1-(piperidin-4-ylcarbonyl)-thiosemicarbazide hydrochloride (1)-a potent topoisomerase II inhibitor-was measured using a MTT assay. It was found that the compound decreased the number of viable cells in both estrogen receptor-positive MCF-7 and estrogen receptor-negative MDA-MB-231breast cancer cells
Juan Liu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 93, 245-249 (2012-04-10)
A simple and highly selective colorimetric sensor (L1) bearing thiosemicarbazide moiety as binding site and nitrophenyl moiety as signal group were synthesized. Sensor L1 showed great colorimetric single selectivity and high sensitivity for mercury cation in DMSO and DMSO/H(2)O binary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service