Skip to Content
Merck
All Photos(3)

Key Documents

861340

Sigma-Aldrich

Thionin acetate salt

certified by the Biological Stain Commission, Dye content ≥85 %

Synonym(s):

3,7-Diamino-5-phenothiazinium acetate, Lauth’s violet

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9N3S · C2H4O2
CAS Number:
Molecular Weight:
287.34
Colour Index Number:
52000
Beilstein:
4345073
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content, ≥85%

technique(s)

microbe id | staining: suitable

λmax

598 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CC([O-])=O.Nc1ccc2nc3ccc(N)cc3[s+]c2c1

InChI

1S/C12H10N3S.C2H4O2/c13-7-1-3-9-11(5-7)16-12-6-8(14)2-4-10(12)15-9;1-2(3)4/h1-6H,13-14H2;1H3,(H,3,4)/q+1;/p-1

InChI key

OWXBIRAFHWASMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Thionin is a diaminothiazine, metachromatic cationic dye.

Application

Thionin is widely used as a biological stain, especially in histology. It has been used in the Nissl stain for the labeling of neuronal cell bodies.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cerebellar Purkinje cell neurodegeneration after cardiac arrest: effect of therapeutic hypothermia.
Paine MG, et al.
Resuscitation, 83, 1511-1511 (2012)
Contribution of carbohydrate histochemistry to glycobiology.
Danguy A, et al.
Histology and Histopathology, 9, 155-155 (1994)
Selvaraj Baskar et al.
Biosensors & bioelectronics, 33(1), 95-99 (2012-01-24)
We report here a novel detection scheme for simultaneous detection of NADH and H(2)O(2) based on a bifunctional poly(thionine)-modified electrode. Electropolymerization of thionine on a "preanodized" screen-printed carbon electrode effectively lowers the oxidation potential of NADH to 0.15 V (vs.
Zhoumin Sun et al.
Analytica chimica acta, 761, 84-91 (2013-01-15)
In this paper, we fabricate a sensitive and stable amperometric UA amperometric biosensor using nanobiocomposite derived from thionine modified graphene oxide in this study. A simple wet-chemical strategy for synthesis of thionine-graphene oxide hybrid nanosheets (T-GOs) through π-π stacking has
Zhanxia Zhang et al.
Journal of the American Chemical Society, 135(5), 1934-1940 (2013-01-10)
The fluorescent dyes methylene blue, MB(+), and thionine, Th(+), can be trapped in the pores of mesoporous silica, MP-SiO(2), by means of functional nanostructures consisting of the Mg(2+)- or Zn(2+)-dependent DNAzyme sequences. In the presence of Mg(2+) or Zn(2+) ions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service