Skip to Content
Merck
All Photos(1)

Key Documents

78513

Supelco

D-Mannitol

≥99.9999% (metals basis), for boron determination

Synonym(s):

Mannite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721898
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (sum of enantiomers, HPLC)
≥99.9999% (metals basis)

form

powder

optical activity

[α]/D +24.0±1° in borax solution acc. to ACS

mp

166-169 °C
167-170 °C (lit.)

anion traces

bromide (Br-): ≤1000 μg/kg
chloride (Cl-): ≤1000 μg/kg
fluoride (F-): ≤1000 μg/kg
nitrate (NO3-): ≤1000 μg/kg
nitrite (NO2-): ≤1000 μg/kg
phosphate (PO43-): ≤1000 μg/kg
sulfate (SO42-): ≤1000 μg/kg

cation traces

Ag: ≤0.01 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.01 mg/kg
Au: ≤0.01 mg/kg
B: ≤0.05 mg/kg
Ba: ≤0.05 mg/kg
Be: ≤0.01 mg/kg
Bi: ≤0.01 mg/kg
Ca: ≤0.2 mg/kg
Cd: ≤0.01 mg/kg
Ce: ≤0.01 mg/kg
Co: ≤0.01 mg/kg
Cr: ≤0.01 mg/kg
Cs: ≤0.01 mg/kg
Cu: ≤0.01 mg/kg
Dy: ≤0.01 mg/kg
Er: ≤0.01 mg/kg
Eu: ≤0.01 mg/kg
Fe: ≤0.05 mg/kg
Ga: ≤0.01 mg/kg
Gd: ≤0.01 mg/kg
Ge: ≤0.01 mg/kg
Hf: ≤0.01 mg/kg
Hg: ≤0.001 mg/kg
Ho: ≤0.01 mg/kg
In: ≤0.01 mg/kg
Ir: ≤0.01 mg/kg
K: ≤0.1 mg/kg
La: ≤0.01 mg/kg
Li: ≤0.01 mg/kg
Lu: ≤0.01 mg/kg
Mg: ≤0.05 mg/kg
Mn: ≤0.01 mg/kg
Mo: ≤0.01 mg/kg
Na: ≤0.5 mg/kg
Nb: ≤0.01 mg/kg
Nd: ≤0.01 mg/kg
Ni: ≤0.01 mg/kg
Os: ≤0.01 mg/kg
Pb: ≤0.01 mg/kg
Pd: ≤0.01 mg/kg
Pr: ≤0.01 mg/kg
Pt: ≤0.01 mg/kg
Rb: ≤0.01 mg/kg
Re: ≤0.01 mg/kg
Rh: ≤0.01 mg/kg
Ru: ≤0.01 mg/kg
Sb: ≤0.01 mg/kg
Sc: ≤0.05 mg/kg
Se: ≤0.01 mg/kg
Sm: ≤0.01 mg/kg
Sn: ≤0.01 mg/kg
Sr: ≤0.01 mg/kg
Ta: ≤0.01 mg/kg
Tb: ≤0.01 mg/kg
Te: ≤0.01 mg/kg
Th: ≤0.01 mg/kg
Ti: ≤0.05 mg/kg
Tl: ≤0.01 mg/kg
Tm: ≤0.01 mg/kg
U: ≤0.01 mg/kg
V: ≤0.01 mg/kg
W: ≤0.01 mg/kg
Y: ≤0.01 mg/kg
Yb: ≤0.01 mg/kg
Zn: ≤0.01 mg/kg
Zr: ≤0.01 mg/kg

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4-,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-KVTDHHQDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

D-Mannitol may be used as a reference standard in the determination of D-mannitol in confectioneries using high-performance liquid chromatography (HPLC).[1]

Biochem/physiol Actions

A sugar alcohol sweet tastant. Used in sweetness inhibition studies.

Linkage

Visit the IC Portal to learn more

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S Nojiri et al.
Journal of chromatography. A, 893(1), 195-200 (2000-10-24)
A method was developed for the determination of sugar alcohols, meso-erythritol, xylitol, D-glucitol, D-mannitol, maltitol and parachinit by high-performance liquid chromatography (HPLC). The sugar alcohols were converted into strong ultraviolet (UV)-absorbing derivatives with p-nitrobenzoyl chloride. HPLC was performed on a
Heikki O Koskela et al.
Chest, 124(4), 1341-1349 (2003-10-14)
To compare the sensitivity and validity of mannitol, histamine, and cold air challenges to demonstrate the effect of inhaled corticosteroids (ICS) in asthma. A prospective study. Seventeen patients with recently diagnosed, steroid-naive asthma who fulfilled the diagnostic criteria of Finnish
Heikki O Koskela et al.
Chest, 125(6), 1985-1992 (2004-06-11)
To define whether coughing during mannitol challenge is a nonspecific side effect of this challenge or is associated with asthma. A prospective study. University hospital. Thirty-seven steroid-naive, asthmatic subjects and 10 healthy subjects. The participants completed a symptom questionnaire, recorded
Diana Bilton et al.
Thorax, 69(12), 1073-1079 (2014-09-24)
Bronchiectasis is characterised by excessive production of mucus and pulmonary exacerbations. Inhaled osmotic agents may enhance mucociliary clearance, but few long-term clinical trials have been conducted. To determine the impact of inhaled mannitol on exacerbation rates in patients with non-cystic
Traumatic intracranial hypertension.
Nino Stocchetti et al.
The New England journal of medicine, 370(22), 2121-2130 (2014-05-30)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service