Skip to Content
Merck
All Photos(1)

Key Documents

76307

Supelco

Pefabloc® SC

analytical standard

Synonym(s):

4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, AEBSF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10FNO2S · HCl
CAS Number:
Molecular Weight:
239.69
Beilstein:
7604627
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

form

powder or crystals

shelf life

5 yr

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

FS(C1=CC=C(CCN)C=C1)(=O)=O.[H]Cl

InChI

1S/C8H10FNO2S.ClH/c9-13(11,12)8-3-1-7(2-4-8)5-6-10;/h1-4H,5-6,10H2;1H

InChI key

WRDABNWSWOHGMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pefabloc® SC is a potent, nontoxic and relatively easy to use, irreversible serine proteinase inhibitor. It is reported to efficiently inhibit the platelet activating factor (PAF)-degrading acetylhydrolase (acetylhydrolase) in both humans and rats.
Pefabloc® SC is a trypsin inhibitor, which can be used in the synthesis of its arginine-like analogue, 4-(2-guanidinoethyl)benzenesulfonyl fluoride.

Application

Pefabloc® SC may be used as an analytical standard in determining the native oligomeric state and substrate specificity of rat NTPDase1 and NTPDase2 after heterologous expression in Xenopus oocytes. It may also be used as an esterase inhibitor, which is added to the whole blood to improve the stability of ciclesonide for the determination of ciclesonide (CIC) and its active metabolite, desisobutyryl-ciclesonide (des-CIC), in human plasma using liquid chromatography-atmospheric pressure chemical ionization-tandem mass spectrometric (LC-APCI-MS/MS) method.
Recommended concentrations for use are 0.1-1 mM.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride (AEBSF) is an irreversible serine protease inhibitor. Inhibition constants are similar to those of phenylmethanesulfonyl fluoride (PMSF) and diisopropylfluorophosphate (DFP). AEBSF has been shown to inhibit trypsin, chymotrypsin, plasmin kallikrein, and thrombin. As an alternative to PMSF and DFP, AEBSF offers lower toxicity, improved solubility in water, and improved stability in aqueous solutions. AEBSF can be used in cell culture in concentrations of up to 0.25 mM.

Other Notes

Potent and irreversible inhibitor of serine proteases; in contrast to PMSF, it is water soluble and non-toxic; Unaffected by albumin

Legal Information

Pefabloc is a registered trademark of Pentapharm

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A trypsin-based bistable switch
Postma.J.GS, et al.
Tetrahedron (2017)
Simultaneous determination of ciclesonide and its active metabolite desisobutyryl-ciclesonide in human plasma by LC?APCI-MS/MS: Application to pharmacokinetic study in healthy Chinese volunteer
Su X-M, et al.
Journal of Pharmaceutical and Biomedical Analysis, 55(1), 230-235 (2011)
Pefabloc, 4-[2-aminoethyl] benzenesulfonyl fluoride, is a new, potent nontoxic and irreversible inhibitor of PAF-degrading acetylhydrolase
Dentan C, et al.
Biochimica et Biophysica Acta, 1299(3), 353-357 (1996)
Determination of native oligomeric state and substrate specificity of rat NTPDase1 and NTPDase2 after heterologous expression in Xenopus oocytes
Failer UB, et al.
European Journal of Biochemistry, 270(8), 1802-1809 (2003)
W B Lawson et al.
Folia haematologica (Leipzig, Germany : 1928), 109(1), 52-60 (1982-01-01)
The effects of blood plasma and some plasma constituents on several types of thrombin inhibitors were quite varied. Two active esters were rapidly destroyed by serum albumin; one of these reacted initially with Lys-199, the residue that is also acylated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service