Skip to Content
Merck
All Photos(1)

Key Documents

P5318

Sigma-Aldrich

Pepstatin A

powder, ≥90% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C34H63N5O9
CAS Number:
Molecular Weight:
685.89
Beilstein:
2201362
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Product Name

Pepstatin A, microbial, ≥90% (HPLC)

biological source

microbial

Quality Level

Assay

≥90% (HPLC)

form

powder

mp

233 °C (dec.) (lit.)

solubility

ethanol: 1-2 mg/mL (with heat up to 60 °C)

storage temp.

2-8°C

SMILES string

CC(C)C[C@H](NC(=O)[C@H](C)NC(=O)C[C@H](O)[C@H](CC(C)C)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CC(C)C)C(C)C)C(C)C)[C@@H](O)CC(O)=O

InChI

1S/C34H63N5O9/c1-17(2)12-23(37-33(47)31(21(9)10)39-34(48)30(20(7)8)38-27(42)14-19(5)6)25(40)15-28(43)35-22(11)32(46)36-24(13-18(3)4)26(41)16-29(44)45/h17-26,30-31,40-41H,12-16H2,1-11H3,(H,35,43)(H,36,46)(H,37,47)(H,38,42)(H,39,48)(H,44,45)/t22-,23-,24-,25-,26-,30-,31-/m0/s1

InChI key

FAXGPCHRFPCXOO-LXTPJMTPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pepstatin A is a naturally occurring chemotactic peptide and inhibitor of aspartic proteases that was initially isolated from culture filtrates of various Actinomycetes species, with the initial name simply of “pepstatin”. This name was later modified to “pepstatin A” to distinguish the original pepstatin from later derivatives. Pepstatin A notably contains the unusual amino acid 4-amino-hydroxy-6-methylheptanoic acid (AHMHA), which is known also as statine. The amino acid sequence of pepstatin A is N-Isovaleryl-L-Valyl-L-Valyl-AHMHA-L-Alanyl-AHMHA.

Application

Can be used in conjunction with E64-d and Leupeptin A to inhibit the degradation of autophagic cargo inside autophagosomes. For this application, the working concentration is typically between 1-10 μM.

Biochem/physiol Actions

Pepstatin A is an inhibitor of acid proteases (aspartyl peptidases). It forms a 1:1 complex with proteases such as pepsin, renin, cathepsin D, bovine chymosin, and protease B (Aspergillus niger). The inhibitor is highly selective and does not inhibit thiol proteases, neutral proteases, or serine proteases. Solubilized γ-secretase and retroviral protease are also inhibited by Pepstatin A. It has been used to characterize proteases from several sources.
Potent inhibitor of acid proteases, including pepsin, renin and cathepsin D and many microbial aspartic proteases. Effective concentration 1 μM. Stable for about 1 day.

Preparation Note

Pepstatin can be dissolved at 1 mg/ml in 10% (v/v) acetic acid in methanol (9:1 methanol:acetic acid). The inclusion of acetic acid may be necessary to dissolve this peptide in methanol or DMSO. It has been dissolved at 1 to 2 mg/ml in ethanol, but heat may be required for complete dissolution. Solutions of Pepstatin A can be heated as high as 60 °C without any decomposition of the peptide.
Stock solutions at 1 mg/ml are stable at least a week at 4 °C. A 1 mM solution in methanol or DMSO is stable for months at -20 °C. If solutions become darker yellow, the reagent is hydrolyzing.
A working concentration of 1 μM is stable for at least one day at room temperature.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaodong Li et al.
Investigative ophthalmology & visual science, 51(10), 5019-5029 (2010-05-14)
Lacrimal glands (LGs) of male NOD mice, a model of Sjögren's syndrome (SjS), exhibit immune cell infiltration and lipid deposition. The mechanism of SjS was further investigated by characterizing gene expression profiles of NOD mouse LGs in comparison with those
Rosa Berga-Bolaños et al.
Journal of immunology (Baltimore, Md. : 1950), 185(11), 6624-6635 (2010-11-03)
Immune cells rely on the transcription factor NFAT5 to adapt to hypertonic stress. The hypertonicity-dependent role of NFAT5 in T cells in vivo remains unclear because mouse models of NFAT5 deficiency have produced substantially different T cell phenotypes. In this
Satoshi Nakamura et al.
Scientific reports, 10(1), 12242-12242 (2020-07-24)
Vitamin D deficiency is a recognized risk factor for sarcopenia development, but mechanisms underlying this outcome are unclear. Here, we show that low vitamin D status worsens immobilization-induced muscle atrophy in mice. Mice globally lacking vitamin D receptor (VDR) exhibited
Weizhong Chang et al.
Human molecular genetics, 19(2), 223-234 (2009-10-23)
Null mutations in cartilage-associated protein (CRTAP) and prolyl 3-hydroxylase 1 (P3H1/LEPRE1) cause types VII and VIII OI, respectively, two novel recessive forms of osteogenesis imperfecta (OI) with severe to lethal bone dysplasia and overmodification of the type I collagen helical
Pearl V Ryder et al.
The Journal of cell biology, 219(12) (2020-11-17)
Centrosomes are microtubule-organizing centers required for error-free mitosis and embryonic development. The microtubule-nucleating activity of centrosomes is conferred by the pericentriolar material (PCM), a composite of numerous proteins subject to cell cycle-dependent oscillations in levels and organization. In diverse cell

Articles

ReadyShield® phosphatase and protease inhibitor cocktail FAQ for sample protection in a variety of cell types and tissue extracts, including mammalian, plant, and microbial samples. Our ReadyShield® Protease Inhibitor Cocktail is a non-freezing solution that contains inhibitors with a broad specificity for serine, cysteine, acid proteases and aminopeptidases.

ReadyShield® phosphatase and protease inhibitor cocktail FAQ for sample protection in a variety of cell types and tissue extracts, including mammalian, plant, and microbial samples. Our ReadyShield® Protease Inhibitor Cocktail is a non-freezing solution that contains inhibitors with a broad specificity for serine, cysteine, acid proteases and aminopeptidases.

ReadyShield® phosphatase and protease inhibitor cocktail FAQ for sample protection in a variety of cell types and tissue extracts, including mammalian, plant, and microbial samples. Our ReadyShield® Protease Inhibitor Cocktail is a non-freezing solution that contains inhibitors with a broad specificity for serine, cysteine, acid proteases and aminopeptidases.

ReadyShield® phosphatase and protease inhibitor cocktail FAQ for sample protection in a variety of cell types and tissue extracts, including mammalian, plant, and microbial samples. Our ReadyShield® Protease Inhibitor Cocktail is a non-freezing solution that contains inhibitors with a broad specificity for serine, cysteine, acid proteases and aminopeptidases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service