Skip to Content
Merck
All Photos(1)

Key Documents

73960

Sigma-Aldrich

D-Galacturonic acid sodium salt

≥95.0% (T)

Synonym(s):

Sodium D-galacturonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9NaO7
CAS Number:
Molecular Weight:
216.12
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

biological source

plant

Quality Level

Assay

≥95.0% (T)

form

powder or crystals

optical activity

[α]/D +36.5±2°, 5 hr, c = 10% in H2O

SMILES string

[Na+].OC1O[C@@H]([C@H](O)[C@H](O)[C@H]1O)C([O-])=O

InChI

1S/C6H10O7.Na/c7-1-2(8)4(5(10)11)13-6(12)3(1)9;/h1-4,6-9,12H,(H,10,11);/q;+1/p-1/t1-,2+,3+,4-,6?;/m0./s1

InChI key

MSXHSNHNTORCAW-KSSASCOMSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chatchaya Onumpai et al.
Applied and environmental microbiology, 77(16), 5747-5754 (2011-07-05)
To evaluate the fermentation properties of oligosaccharides derived from pectins and their parent polysaccharides, a 5-ml-working-volume, pH- and temperature-controlled fermentor was tested. Six pectic oligosaccharides representing specific substructures found within pectins were prepared. These consisted of oligogalacturonides (average degrees of
S Rasul et al.
Plant, cell & environment, 35(8), 1483-1499 (2012-03-08)
Nitric oxide (NO) regulates a wide range of plant processes from development to environmental adaptation. In this study, we investigated the production and/or function of NO in Arabidopsis thaliana leaf discs and plants elicited by oligogalacturonides (OGs) and challenged with
Joosu Kuivanen et al.
Applied and environmental microbiology, 78(24), 8676-8683 (2012-10-09)
D-Galacturonic acid, the main monomer of pectin, is an attractive substrate for bioconversions, since pectin-rich biomass is abundantly available and pectin is easily hydrolyzed. l-Galactonic acid is an intermediate in the eukaryotic pathway for d-galacturonic acid catabolism, but extracellular accumulation
Eline H Huisjes et al.
Applied and environmental microbiology, 78(15), 5052-5059 (2012-05-15)
The efficient fermentation of mixed substrates is essential for the microbial conversion of second-generation feedstocks, including pectin-rich waste streams such as citrus peel and sugar beet pulp. Galacturonic acid is a major constituent of hydrolysates of these pectin-rich materials. The
Adebanjo Ayobamidele Badejo et al.
Journal of experimental botany, 63(1), 229-239 (2011-10-11)
The D-mannose/L-galactose pathway for the biosynthesis of vitamin C (L-ascorbic acid; AsA) has greatly improved the understanding of this indispensable compound in plants, where it plays multifunctional roles. However, it is yet to be proven whether the same pathway holds

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service