Skip to Content
Merck
All Photos(1)

Documents

72999

Supelco

Viridiflorol

analytical standard

Synonym(s):

(1aR,4S,4aS,7R,7aS)-1,1,4,7-Tetramethyl-2,3,4a,5,6,7,7a,7b-octahydro-1aH-cyclopropa[e]azulen-4-ol, [1aR-(1aα,4β,4aβ,7α,7aβ,7bα)]-Decahydro-1,1,4,7-tetramethyl-1H-cycloprop[e]azulen-4-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC1(C)[C@@H]2[C@H]1CC[C@@](O)(C)[C@]3([H])[C@@]2([H])[C@H](C)CC3

InChI

1S/C15H26O/c1-9-5-6-10-12(9)13-11(14(13,2)3)7-8-15(10,4)16/h9-13,16H,5-8H2,1-4H3/t9-,10+,11-,12-,13-,15+/m1/s1

InChI key

AYXPYQRXGNDJFU-IMNVLQEYSA-N

General description

Viridiflorol belongs to the class of aromadendrene sesquiterpenoids, isolated from the leaves of Salvia officinalis and detected in leaves of Salvia triloba.

Application

Viridiflorol may be used as a reference standard in the determination of viridiflorol in different types of Chinese eaglewood using gas chromatography coupled with mass spectrometry (GC-MS).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GC-MS analysis of volatile constituents from five different kinds of Chinese eaglewood.
LWM, et al.
Zhong Yao Cai = ZhongYaoCai = Journal of Chinese Medicinal Materials, 30(5), 551-555 (2007)
Biomimetic Synthesis of (+)-Ledene,(+)-Viridiflorol,(-)-Palustrol,(+)-Spathulenol, and Psiguadial A, C, and D via the Platform Terpene (+)-Bicyclogermacrene.
Tran NC and Cramer N
Chemistry?A European Journal , 20(34), 10654-10660 (2014)
The presence of viridiflorol in Salvia officinalis.
Karl C, et al.
Planta Medica, 44(03), 188-189 (1982)
Nurhayat Tabanca et al.
Biomolecules, 10(5) (2020-05-02)
The Mediterranean fruit fly, Ceratitis capitata (Wiedemann) (Diptera: Tephritidae) poses a major threat to fruit and vegetable production in the United States and throughout the world. New attractants and detection methods could improve control strategies for this invasive pest. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service