Skip to Content
Merck
All Photos(1)

Key Documents

72020

Sigma-Aldrich

Sodium tetraphenylborate

puriss. p.a., ACS reagent, ≥99.5% (NT)

Synonym(s):

Tetraphenylboron sodium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)4BNa
CAS Number:
Molecular Weight:
342.22
Beilstein:
3599783
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.5% (NT)
form:
solid
Pricing and availability is not currently available.

grade

ACS reagent
puriss. p.a.

Quality Level

Assay

≥99.5% (NT)

form

solid

loss

≤0.5% loss on drying, 110 °C

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤100 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤500 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[Na+].c1ccc(cc1)[B-](c2ccccc2)(c3ccccc3)c4ccccc4

InChI

1S/C24H20B.Na/c1-5-13-21(14-6-1)25(22-15-7-2-8-16-22,23-17-9-3-10-18-23)24-19-11-4-12-20-24;/h1-20H;/q-1;+1

InChI key

HFSRCEJMTLMDLI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sodium tetraphenylborate (Ph4BNa) undergoes rhodium-catalyzed addition reactions with N-phenylsulfonyl aldimines to afford R(Ph)CHNHSO2Ph.[1]

Application

Sodium tetraphenylborate (Ph4BNa) may be used in the synthesis of polyfunctional biaryls via modified Suzuki cross-coupling reaction.[2] It may be employed as a derivatization reagent for the analysis of methylmercury and ethylmercury by gas chromatography (GC) coupled with atomic fluorescence spectrometry (AFS), microwave-induced plasma atomic emission spectrometry (AES) and mass spectrometry (MS).[3]

Other Notes

Reagent for the precipitation and titration of potassium[4][5]; For the amperometric titration of amines, quaternary ammonium compounds and potassium[6]; For the titration of quaternary ammonium surfactants[7]; Titrant for amines and NH4Cl in the presence of a K selective electrode[8]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recorded amperometric tetraphenylborate titrations of amines, quaternary nitrogen compounds, and potassium salts.
J E Sinsheimer et al.
Journal of pharmaceutical sciences, 54(5), 805-806 (1965-05-01)
K. Vytras
Collection of Czechoslovak Chemical Communications, 42, 3168-3168 (1977)
Y Cai et al.
Journal of chromatography. A, 876(1-2), 147-155 (2000-05-24)
Several hyphenated analytical techniques, including gas chromatography (GC) coupled with atomic fluorescence spectrometry (AFS), microwave-induced plasma atomic emission spectrometry (AES), and mass spectrometry (MS), have been evaluated for methylmercury and ethylmercury analysis following aqueous derivatization with both sodium tetraethylborate and
Ligandless palladium catalyzed reactions of arylboronic acids and sodium tetraphenylborate with aryl halides in aqueous media.
Bumagin NA and Bykov VV.
Tetrahedron, 53(42), 14437-14450 (1997)
J M Skinner et al.
Talanta, 14(12), 1393-1401 (1967-12-01)
An automatic ultraviolet absorptiometric method has been developed for the determination of potassium. The method involves precipitation of potassium by addition of a known excess of sodium tetraphenylborate, removal of the potassium tetraphenylborate precipitate by filtration, and measurement of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service