Skip to Content
Merck
All Photos(1)

Key Documents

71480

Supelco

Sodium diethyldithiocarbamate trihydrate

ACS reagent, for the determination and separation of heavy metals, ≥97%

Synonym(s):

Cupral, Diethyldithiocarbamic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NCSSNa · 3H2O
CAS Number:
Molecular Weight:
225.31
Beilstein:
3920507
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

5.9 (vs air)

Assay

≥97%

form

powder

quality

for the determination and separation of heavy metals

mp

95-98.5 °C (lit.)

cation traces

Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
Mn: ≤5 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

[H]O[H].[H]O[H].[H]O[H].CCN(CC)C(=S)S[Na]

InChI

1S/C5H11NS2.Na.3H2O/c1-3-6(4-2)5(7)8;;;;/h3-4H2,1-2H3,(H,7,8);;3*1H2/q;+1;;;/p-1

InChI key

WWGXHTXOZKVJDN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium diethyldithiocarbamate trihydrate is a chelating agent, generally used in mobilizing toxic metals from the human tissues and experimental animals. It exists as a white or colorless solid, and is freely soluble with gradual decomposition.

Application

Sodium diethyldithiocarbamate trihydrate was used during the synthesis of N-allylpeptides, in a study outlining the strategy to synthesize and purify a hydrogen bond surrogate (HBS) α-helix in ∼1 week.
Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues.
Spin trap used in conjunction with Fe2+ to detect NO in brain, kidney, liver, and other tissues. Simultaneous direct measurement of NO* and pO2 has been reported. A copper chelator, it has been used to quantitate copper in biological materials by ESR and is an inhibitor of superoxide dismutase, ascorbate oxidase, and other enzymes.

Biochem/physiol Actions

Inhibits induction of macrophage nitric oxide synthase.

Other Notes

Reagent used for the extraction of trace metals into organic solvents for atomic absorption spectroscopy; Precipitation of trace metals for x-ray fluorescence spectroscopy

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.L. Cheng et al.
Handbook of Organic Analytical Reagents (1982)
Inhibition of induction of benzpyrene hydroxylase by nickel carbonyl.
Sunderman, F. William.
Cancer Research, 27.5, 950-955 (1967)
María N Oviedo et al.
Analytical methods : advancing methods and applications, 13(8), 1033-1042 (2021-02-09)
Efficient separation and preconcentration of inorganic Sb species in different water samples were performed in this work by a novel dispersive liquid-liquid microextraction (DLLME) method based on the application of a magnetic ionic liquid (MIL) and electrothermal atomic absorption spectroscopy
S. Bruggerhoff et al.
Fresenius Journal of Analytical Chemistry, 311, 252-252 (1982)
Paul A. Bouis
Reagent Chemicals: Specifications and Procedures, 609-609 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service