Skip to Content
Merck
All Photos(1)

Key Documents

742546

Sigma-Aldrich

Hydrogen sulfide solution

0.8 M in THF

Synonym(s):

Dihydrogen monosulfide, Hydrosulfuric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H2S
CAS Number:
Molecular Weight:
34.08
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

0.79 M in THF

form

liquid

concentration

0.7-0.9 M in THF

suitability

conforms to structure for Proton NMR spectrum

storage temp.

2-8°C

SMILES string

S

InChI

1S/H2S/h1H2

InChI key

RWSOTUBLDIXVET-UHFFFAOYSA-N

Application

Hydrogen sulfide solution can be used in the preparation of lead sulfide (PbS), titanium disulfide(TiS2) and cadmium sulfide (CdS).[1][2][3] It is also used as a ligand in the synthesis of ammineruthenium complexes.[4]

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup - Solvent

Flash Point(C)

-17 °C - closed cup - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Modified Sol-Gel Processing for Titanium Disulfide.
Go, Yong Bok et al.
J. Korean Chem. Soc., 41(3), 130-137 (1997)
Cadmium sulphide quantum dots in morphologically tunable triblock copolymer aggregates.
Duxin, Nicolas et al.
Journal of the American Chemical Society, 127(28), 10063-10069 (2005)
Amine Ruthenium complexes of hydrogen sulfide and related sulfur ligands.
Kuehn CC
Journal of the American Chemical Society, 98(3), 689-702 (1976)
Size variation of PbS particles in high-refractive-index nanocomposites.
Kyprianidou-Leodidou, Tasoula et al.
The Journal of Physical Chemistry, 98(36), 8992-8997 (1994)
A Livermore et al.
Electroencephalography and clinical neurophysiology, 83(3), 201-210 (1992-09-01)
The aim of the study was to investigate the interaction of the olfactory and somatosensory systems in the perception of chemical stimuli. Stimuli were chosen so as to selectively activate the olfactory (hydrogen sulphide, H2S) and trigeminal (carbon dioxide, CO2)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service