Skip to Content
Merck
All Photos(1)

Key Documents

67414

Sigma-Aldrich

Sodium fluoride 0.5 M solution

Synonym(s):

Sodium fluoride solution, Additive Screening Solution 19/Kit-No 78374

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaF
CAS Number:
Molecular Weight:
41.99
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.32
grade:
for analytical purposes
Pricing and availability is not currently available.

grade

for analytical purposes

Quality Level

density

1.02 g/mL at 20 °C
1.005 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[F-].[Na+]

InChI

1S/FH.Na/h1H;/q;+1/p-1

InChI key

PUZPDOWCWNUUKD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium fluoride (NaF) has been used as a component:

  • of the extraction buffer to extract proteins from Nicotiana benthamiana leaf[1]
  • of the mammalian tissue lysis buffer for homogenization of mice brain[2]
  • of the lysis buffer for lysing Caenorhabditis elegans to extract proteins[3]

Biochem/physiol Actions

Sodium fluoride (NaF) is used as a major constituent in toothpaste, mouth rinses, and mouthwashes.[4] It is used as a pesticide to control larvae and insects in barns, warehouses, houses, and storage areas.[5] NaF also acts as an esterase inhibitor.[6]

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miscellaneous Inorganic Toxicants
Cope R
Archives of Veterinary Science, 289-332 (2017)
Jamal Ahmad Ansari et al.
Environmental toxicology and pharmacology, 92, 103858-103858 (2022-03-31)
Arsenic activates microglia and exerts bystander effects on neuron. The present study is focused to test whether minocycline, a second generation antibiotic, can reverse the effect of developmental arsenic exposure on microglial activation and function. Pregnant Balb/c dams were gavaged
Cyril Statzer et al.
Nature communications, 13(1), 967-967 (2022-02-20)
Inhibition of the master growth regulator mTORC1 (mechanistic target of rapamycin complex 1) slows ageing across phyla, in part by reducing protein synthesis. Various stresses globally suppress protein synthesis through the integrated stress response (ISR), resulting in preferential translation of
Laïla Giordano et al.
iScience, 25(3), 103877-103877 (2022-03-05)
Malectins from the oligosaccharyltransferase (OST) complex in the endoplasmic reticulum (ER) of animal cells are involved in ER quality control and contribute to the Unfolded Protein Response (UPR). Malectins are not found in plant cells, but malectin-like domains (MLDs) are
Melissa van Kranenburg et al.
Human mutation, 36(2), 200-209 (2014-11-05)
The mechanistic target of rapamycin complex 1 (TORC1) senses nutrient availability to regulate eukaryotic anabolic metabolism. In response to limiting concentrations of amino acids, TORC1 kinase activity is inhibited through the GATOR-1 complex. Mutations in DEPDC5, that encodes one of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service