Skip to Content
Merck
All Photos(1)

Key Documents

59510

Supelco

Methanesulfonic acid

suitable for HPLC, LiChropur, ≥99.5% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO3H
CAS Number:
Molecular Weight:
96.11
Beilstein:
1446024
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NB.21

vapor density

3.3 (vs air)

Quality Level

vapor pressure

1 mmHg ( 20 °C)

Assay

≥99.5% (T)

form

liquid

quality

LiChropur

technique(s)

HPLC: suitable

color

APHA: ≤35

refractive index

n20/D 1.428-1.430
n20/D 1.429 (lit.)

bp

167 °C/10 mmHg (lit.)

mp

17-19 °C (lit.)

density

1.481 g/mL at 25 °C (lit.)

suitability

corresponds to standard for HPLC

SMILES string

O=S(O)(C)=O

InChI

1S/CH4O3S/c1-5(2,3)4/h1H3,(H,2,3,4)

InChI key

AFVFQIVMOAPDHO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Analysis Note

test by UV spectroscopy corresponds to requirements

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

372.2 °F - closed cup

Flash Point(C)

189 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Mohammad M Fares et al.
Biomaterials science, 6(11), 2938-2950 (2018-09-25)
The design of new hydrogel-based biomaterials with tunable physical and biological properties is essential for the advancement of applications related to tissue engineering and regenerative medicine. For instance, interpenetrating polymer network (IPN) and semi-IPN hydrogels have been widely explored to
Adnan H Mahmood et al.
International journal of pharmaceutics, 481(1-2), 71-78 (2015-02-01)
To introduce better antibiotics for the treatment of some infectious diseases in sheep and to expand the range of antibiotics available for veterinary medicine, pharmacokinetics of two antibiotics marbofloxacin (MBX) and trovafloxacin (TVX) were investigated in sheep after intramuscular injection.
Chris Twelves et al.
Breast cancer research and treatment, 148(3), 553-561 (2014-11-09)
Data from two phase 3 studies of eribulin were pooled in analyses initially requested by the European Medicines Agency to assess whether specific patient subgroups, previously treated with an anthracycline and a taxane, benefited from eribulin. Study 305/EMBRACE included women
Zhan Zhang et al.
Journal of the American Chemical Society, 136(21), 7591-7594 (2014-05-13)
A large porphyrin analogue, cyclo[6]pyridine[6]pyrrole, containing no meso bridging atoms, has been synthesized through Suzuki coupling. In its neutral form, this macrocycle exists as a mixture of two figure-eight conformers that undergo fast exchange in less polar solvents. Upon protonation
Kay O Johswich et al.
Infection and immunity, 83(1), 354-363 (2014-11-05)
Neisseria meningitidis asymptomatically colonizes the human upper respiratory tract but is also the cause of meningitis and severe septicemia. Carriage or disease evokes an immune response against the infecting strain. Hitherto, we have known little about the breadth of immunity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service