Skip to Content
Merck
All Photos(1)

Documents

5.43838

Supelco

Sodium phosphate dibasic

anhydrous for HPLC LiChropur

Synonym(s):

Sodium phosphate dibasic, di-Sodium hydrogen phosphate anhydrous, sec-Sodium phosphate, Disodium hydrogen phosphate, Disodium phosphate, Sodium hydrogenphosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2HPO4
CAS Number:
Molecular Weight:
141.96
MDL number:
UNSPSC Code:
41116105
EC Index Number:
231-448-7
NACRES:
NB.21

grade

anhydrous

Quality Level

form

solid

technique(s)

HPLC: suitable

pH

8.7-9.3 (20 °C, 10 g/L in H2O)

pKa (25 °C)

(1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)

mp

250 °C

solubility

77 g/L

bulk density

880 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[H]OP([O-])([O-])=O

InChI

1S/2Na.H3O4P/c;;1-5(2,3)4/h;;(H3,1,2,3,4)/q2*+1;/p-2

InChI key

BNIILDVGGAEEIG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Synchronized separation of seven medications representing most commonly prescribed antihypertensive classes by using reversed-phase liquid chromatography: Application for analysis in their combined formulations.: This study utilizes sodium phosphate dibasic in the reversed-phase liquid chromatography process to analyze and separate multiple antihypertensive drugs in combined formulations, enhancing the precision and efficiency of pharmaceutical analysis (Ebeid et al., 2014).

  • [Study of the acupuncture effect on monoamine transmitters in rabbit plasma and brain tissue by high performance liquid chromatography with electrochemical detection].: Sodium phosphate dibasic was used as a buffering agent in the HPLC method to study the effects of acupuncture on monoamine levels in rabbit plasma and brain tissue, providing insights into biochemical responses to traditional Chinese medicine treatments (Hou et al., 2002).

  • The effect of formulation on reduced radioiodide thyroid uptake.: This research explores the role of sodium phosphate dibasic in the formulation of radiopharmaceuticals, specifically its impact on thyroid uptake of radioiodide, with implications for improving diagnostic and therapeutic applications in nuclear medicine (Yu et al., 2002).


Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Separation and identification of oligonucleotides differing by one base in length using Size Exclusion Chromatography combined with multi-angle light scattering (SEC-MALS) detector.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service