Skip to Content
Merck
All Photos(1)

Documents

59060

Supelco

2-Methylbutane

analytical standard

Synonym(s):

Isopentane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)2
CAS Number:
Molecular Weight:
72.15
Beilstein:
1730723
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.6 (vs air)

vapor pressure

11.17 psi ( 20 °C)

Assay

≥99.7% (GC)

autoignition temp.

788 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

8.3 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.354 (lit.)
n20/D 1.354

bp

30 °C (lit.)

density

0.62 g/mL at 25 °C (lit.)

application(s)

petroleum

format

neat

storage temp.

2-8°C

SMILES string

CCC(C)C

InChI

1S/C5H12/c1-4-5(2)3/h5H,4H2,1-3H3

InChI key

QWTDNUCVQCZILF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

2-Methylbutane can undergo dehydrogenation over chromia-alumina catalyst to yield iso-pentene.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-59.8 °F - closed cup

Flash Point(C)

-51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of Isopentane Dehydrogenation on Chromia-Alumina Catalyst with Catalyst Fouling
Noda H, et al.
J. Chem. Eng. Jpn., 7(2), 110-116 (1974)
P Krustrup et al.
British journal of sports medicine, 43(11), 825-831 (2008-12-23)
To examine the effects of regular participation in recreational soccer on health profile, 36 healthy untrained Danish men aged 20-43 years were randomised into a soccer group (SO; n = 13), a running group (RU; n = 12) and a
Benedikt Galliker et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(25), 6161-6168 (2009-05-14)
We have identified two intermediates in the autoxidation of NO*: ONOO*, which was detected by EPR spectroscopy at 295 K and atmospheric pressure in the gas phase, and ONOONO, a red substance produced at 113 K in 2-methylbutane. The red
Craig M Bullock et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(8), 2188-2200 (2014-04-11)
To investigate the role of the sensory neuropeptide calcitonin gene-related peptide (CGRP) in peripheral sensitization in experimental models of osteoarthritis (OA) pain. Experimental knee OA was induced in rats by intraarticular injection of monosodium iodoacetate (MIA) or by transection of
Ryo Ogawa et al.
Development (Cambridge, England), 142(1), 51-61 (2014-12-07)
Muscle satellite cells are indispensable for muscle regeneration, but the functional diversity of their daughter cells is unknown. Here, we show that many Pax7(+)MyoD(-) cells locate both beneath and outside the basal lamina during myofiber maturation. A large majority of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service