Skip to Content
Merck
All Photos(1)

Key Documents

56671

Supelco

(±)-Propylene oxide

analytical standard

Synonym(s):

(±)-Methyloxirane, 1,2-Epoxypropane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O
CAS Number:
Molecular Weight:
58.08
Beilstein:
79763
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2 (vs air)

vapor pressure

29.43 psi ( 55 °C)
8.59 psi ( 20 °C)

Assay

≥99.7% (GC)

autoignition temp.

1378 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

37 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.366 (lit.)
n20/D 1.366

bp

34 °C (lit.)

mp

−112 °C (lit.)

density

0.83 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC1CO1

InChI

1S/C3H6O/c1-3-2-4-3/h3H,2H2,1H3

InChI key

GOOHAUXETOMSMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

(±)-Propylene oxide has been used as a starting material in the preparation of magnetic resonance (MR) contrast agents like Β-EgadMe and Α-EgadMe, which can be used for detecting the presence of Β-galactosidase in vivo.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-36.4 °F - Equilibrium method

Flash Point(C)

-38 °C - Equilibrium method

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lauren M Urbanczyk-Pearson et al.
Nature protocols, 3(3), 341-350 (2008-03-08)
The preparation of beta-EgadMe and alpha-EgadMe, magnetic resonance (MR) contrast agents that can be used for in vivo detection of beta-galactosidase, is described. Diastereomerically pure beta-EgadMe can be synthesized by kinetically resolving the starting material as described in Step 1.
Josep Casamada Ribot et al.
Chemical communications (Cambridge, England), 46(37), 6971-6973 (2010-08-24)
The aqueous gelation of a quaternary ammonium oligo(propylene oxide)-based ionic liquid yields ion gels with a reverse thermoresponsive behavior (i.e., mechanical moduli and viscosity increase with temperature) and enhanced ionic conductivities.
Yuqiang Qian et al.
ACS applied materials & interfaces, 3(9), 3709-3717 (2011-08-23)
Natural vermiculite was modified by cation exchange with long-chain quaternary alkylammonium salts and then dispersed in polyether-based polyols with different structures and ethylene oxide/propylene oxide ratios. The dispersions were evaluated by X-ray scattering and rheology. In all polyol dispersions tested
Javix Thomas et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(16), 4582-4587 (2011-03-25)
Chirality recognition in the hydrogen-bonded glycidol···propylene oxide complex has been studied by using rotational spectroscopy and ab initio calculations. An extensive conformational search has been performed for this binary adduct at the MP2/6-311++G(d,p) level of theory and a total of
Nitai Ecker et al.
Autophagy, 6(7), 879-890 (2010-07-22)
A number of signaling mechanisms have been implicated in the regulation of autophagic trafficking. TOR kinase activity, cAMP levels, and the GAAC pathway have all been suggested to be involved. Here, we closely analyzed the stimuli that underlie induction of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service