Skip to Content
Merck
All Photos(1)

Key Documents

492167

Sigma-Aldrich

D-Glucose-12C6

99.5 atom % 12C

Synonym(s):

Dextrose-12C6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
12C6H12O6
CAS Number:
Molecular Weight:
180.09
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

description

13C-depleted

Quality Level

isotopic purity

99.5 atom % 12C

optical activity

[α]25/D +52.0°, c = 2 in H2O (trace NH4OH)

mp

150-152 °C (lit.)

mass shift

depleted

SMILES string

O[12CH2][12C@H]1O[12CH](O)[12C@H](O)[12C@@H](O)[12C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1/i1+0,2+0,3+0,4+0,5+0,6+0

InChI key

WQZGKKKJIJFFOK-DXVIFZLFSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiali Tang et al.
Nature communications, 10(1), 4057-4057 (2019-09-08)
Simultaneous imaging and treatment of infections remains a major challenge, with most current approaches being effective against only one specific group of bacteria or not being useful for diagnosis. Here we develop multifunctional nanoagents that can potentially be used for
Joanna Potaś et al.
Materials (Basel, Switzerland), 14(1) (2020-12-31)
Polyelectrolyte complexes based on the electrostatic interactions between the polymers mixed are of increasing importance, therefore, the aim of this study was to develop hydrogels composed of anionic tragacanth gum and cationic chitosan with or without the addition of anionic
Yi Xu et al.
Environmental science and pollution research international, 26(9), 9293-9304 (2019-02-07)
In this paper, the effects of cerium oxide nanoparticles (CeO2 NPs) on the group bacterial behaviors were elaborated. After 36-h cultivation, the biofilm biomass was enhanced by the sub-lethal concentrations of 0.5 and 2 mg/L CeO2 NP exposure. Meanwhile, the promoted
Madhavan Chalat et al.
Molecular biology of the cell, 28(3), 452-462 (2016-12-10)
ATP8A2 is a P4-ATPase that flips phosphatidylserine and phosphatidylethanolamine across cell membranes. This generates membrane phospholipid asymmetry, a property important in many cellular processes, including vesicle trafficking. ATP8A2 deficiency causes severe neurodegenerative diseases. We investigated the role of the C-terminus
Sara Groeneboer et al.
Rheumatology (Oxford, England), 50(7), 1226-1235 (2011-02-25)
To compare the ability of different cyclodextrin polysulphate (CDPS) derivatives to affect human articular cartilage cell metabolism in vitro. OA chondrocytes were cultured in alginate and exposed to 5 µg/ml of 2,3,6-tri-O-methyl-β-cyclodextrin (ME-CD), 2,3-di-O-methyl-6-sulphate-β-cyclodextrin (ME-CD-6-S), 2,6-di-O-methyl-3-sulphate-β-cyclodextrin (ME-CD-3-S), (2-carboxyethyl)-β-CDPS (CE-CDPS), (2-hydroxypropyl)-β-CDPS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service