Skip to Content
Merck
All Photos(1)

Documents

46609

Supelco

Penicillin G potassium salt

VETRANAL®, analytical standard

Synonym(s):

Benzylpenicillin potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17KN2O4S
CAS Number:
Molecular Weight:
372.48
Beilstein:
3832841
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
E Number:
E705
NACRES:
NA.24

grade

analytical standard

Quality Level

Agency

EPA 1694

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

antibiotic activity spectrum

Gram-positive bacteria

application(s)

cleaning products
clinical
cosmetics
environmental
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

Mode of action

cell wall synthesis | interferes

storage temp.

2-8°C

SMILES string

[K+].[H][C@]12SC(C)(C)[C@@H](N1C(=O)[C@H]2NC(=O)Cc3ccccc3)C([O-])=O

InChI

1S/C16H18N2O4S.K/c1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);/q;+1/p-1/t11-,12+,14-;/m1./s1

InChI key

IYNDLOXRXUOGIU-LQDWTQKMSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam

Application

Penicillin G is a narrow spectrum antibiotic derived from Streptococcus pneumoniae. It is the drug of choice for groups A, B, C and G streptococci, nonenterococcal group D streptococci, viridians group streptococci, and non-penicillinase producing staphylococcus. The potassium salt has been used to study murosomes of staphylococci and the penicillin-induced lysis of Streptococcus mutans.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Mode of Action: Penicillin G acts by inhibiting cell wall synthesis through binding to penicillin binding proteins (PBPs), inhibiting peptidoglycan chain cross-linking.

Antimicrobial spectrum: This product is active against gram-positive and gram-negative bacteria.

Caution

Solutions should be filter sterilized and stored at 2-8°C for 1 week or at -20°C for more lengthy periods. Solutions are stable at 37°C for 3 days. The sodium salt is soluble in H2O at 100 mg/mL.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vasim Ahmed et al.
Journal of biotechnology, 163(4), 419-424 (2013-01-12)
In the present study, a novel strategy was adopted to synthesize, β lactamase resistant penicillin G molecules by using the unique properties of silver nanoparticles. Ascorbic acid-stabilized spherical monodispersed silver nanoparticles were prepared by a simple chemical reaction. The formation
Tina Lund Leunbach et al.
Ugeskrift for laeger, 175(11), 742-743 (2013-03-14)
A pregnant Caucasian woman with newly diagnosed syphilis in the secondary stage was hospitalized for observation while being treated with penicillin. After one dose of penicillin the foetus was delivered by a caesarean section at gestational age 28 weeks +
Muneki Hotomi et al.
PloS one, 8(3), e58124-e58124 (2013-03-14)
The protection against pneumococcal infections provided by currently available pneumococcal polysaccharide conjugate vaccines are restricted to the limited number of the serotypes included in the vaccine. In the present study, we evaluated the distribution of the pneumococcal capsular type and
Nicolas Dupin et al.
Presse medicale (Paris, France : 1983), 42(4 Pt 1), 446-453 (2013-02-19)
Syphilis is back since 2000. Early syphilis comprises primary syphilis, secondary syphilis and early latent syphilis (less than 1 year duration). During early phases of syphilis, patients are more contagious and neurologic complications are rare. Early neurosyphilis are mostly represented
Eleonor G Lago et al.
Sexually transmitted diseases, 40(2), 85-94 (2013-01-18)
The aim of this study was to investigate clinical features and outcomes of children treated for congenital syphilis (CS). Infants born alive in the public sector of São Lucas Hospital, Porto Alegre, Brazil, 1997 to 2004, whose mothers had syphilis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service